Skip to Content
Merck
All Photos(1)

Key Documents

Z0876

Sigma-Aldrich

trans-Zeatin

BioReagent, suitable for plant cell culture, ≥97%

Synonym(s):

6-(4-Hydroxy-3-methylbut-2-enylamino)purine, N6-(4-Hydroxy-3-methyl-2-buten-1-yl)adenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O
CAS Number:
Molecular Weight:
219.24
Beilstein:
616241
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Assay

≥97%

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

−20°C

SMILES string

C\C(CO)=C/CNc1ncnc2nc[nH]c12

InChI

1S/C10H13N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h2,5-6,16H,3-4H2,1H3,(H2,11,12,13,14,15)/b7-2+

InChI key

UZKQTCBAMSWPJD-FARCUNLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-Zeatin contains a hydroxylated isoprenoid side chain.

Application

trans-Zeatin has been used:
  • as a media supplement for the isolation and culture of pretransitional and transitional embryos
  • as a culture media supplement for incubating alginate strip culture
  • as a growth regulator in modified basal Murashige and Skoog medium

Biochem/physiol Actions

Zeatin upon reduction forms zeatin O-glucoside, which is widely present in plants. It possesses transportation and storage functions, along with defence against zeatin oxidases.
trans-Zeatin is a plant cytokinin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro development of maize immature embryos: a tool for embryogenesis analysis
Matthys-Rochon E, et al.
Journal of Experimental Botany, 49(322), 839-845 (1998)
Isolation of a cytokinin gene, ZOG1, encoding zeatin O-glucosyltransferase from Phaseolus lunatus
Martin RC, et al.
Proceedings of the National Academy of Sciences of the USA, 96(1), 284-289 (1999)
Stoloniferous shoot protoplast, an efficient cell system in potato for somatic cell genetic manipulations
Sharma S, et al.
Sci. Hortic., 128(2), 84-91 (2011)
Silvia Gajdosová et al.
Journal of experimental botany, 62(8), 2827-2840 (2011-02-02)
Cytokinins (CKs) are plant hormones affecting numerous developmental processes. Zeatin and its derivatives are the most important group of isoprenoid CKs. Zeatin occurs as two isomers: while trans-zeatin (transZ) was found to be a bioactive substance, cis-zeatin (cisZ) was reported
Cui Qi et al.
Biosensors & bioelectronics, 41, 157-162 (2012-09-06)
Zeatins, a major type of cytokinin, are ubiquitous in higher plants, and involve in regulating a wide range of developmental processes. The development of highly specific ligands to zeatins would be very useful in plant biological research. Here we describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service