Skip to Content
Merck
All Photos(1)

Key Documents

08535

Sigma-Aldrich

DL-Dihydrozeatin

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O
CAS Number:
Molecular Weight:
221.26
Beilstein:
1215977
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (HPLC)

storage temp.

2-8°C

SMILES string

CC(CO)CCNc1ncnc2nc[nH]c12

InChI

1S/C10H15N5O/c1-7(4-16)2-3-11-9-8-10(13-5-12-8)15-6-14-9/h5-7,16H,2-4H2,1H3,(H2,11,12,13,14,15)

InChI key

XXFACTAYGKKOQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DL-Dihydrozeatin can be converted into zeatin by the enzyme zeatin reductase. It has been identified as a plant growth cytokinin that stimulates flower bud formation.
Metabolite in zeatin biosynthesis, metabolism in radish seedlings, conversion of Zeatin to Dihydrozeatin in Phaseolus Embryos.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Van der Krieken et al.
Plant physiology, 92(3), 565-569 (1990-03-01)
Explants from flower stalks of Nicotiana tabacum L. were cultured on different cytokinins to induce flower bud formation. All cytokinins tested except zeatin and zeatin-riboside induced the same maximal number of flower buds. Benzyladenine, benzyladenosine, and dihydrozeatin were the most
Dihydrozeatin metabolism in radish seedlings.
McGaw, B. A.
Phytochemistry, 23, 1373-1377 (1984)
An Enzyme Mediating the Conversion of Zeatin to Dihydrozeatin in Phaseolus Embryos
Martin, R.
Plant Physiology, 89, 1630-1635 (1990)
R C Martin et al.
Plant physiology, 90(4), 1630-1635 (1989-08-01)
A reductase catalyzing the conversion of zeatin to dihydrozeatin was detected in soluble fractions of immature Phaseolus vulgaris embryos. The enzyme was partially purified by ammonium sulfate fractionation and affinity, gel filtration, and anion exchange chromatography. NADPH was the only
Da Cao et al.
Frontiers in plant science, 11, 605069-605069 (2020-12-18)
Phytohormones are involved in most plant physiological processes and the quantification of endogenous phytohormone levels and related gene expressions is an important approach to studying phytohormone functions. However, the quantification of phytohormones is still challenging due to their extremely low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service