Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C10H13N5
CAS Number:
Molecular Weight:
203.24
NACRES:
NA.72
PubChem Substance ID:
UNSPSC Code:
10171502
MDL number:
product line
BioReagent
Quality Segment
assay
≥98.5%
form
powder
technique(s)
cell culture | plant: suitable
application(s)
agriculture
storage temp.
−20°C
SMILES string
C\C(C)=C\CNc1ncnc2[nH]cnc12
InChI
1S/C10H13N5/c1-7(2)3-4-11-9-8-10(13-5-12-8)15-6-14-9/h3,5-6H,4H2,1-2H3,(H2,11,12,13,14,15)
InChI key
HYVABZIGRDEKCD-UHFFFAOYSA-N
Application
6-(γ,γ-Dimethylallylamino)purine (2iP) is a bacteria-derived riboside cytokinin used to grow plant tissues such as tobacco and soybean callus. 2iP is a precursor of the cytokinin Zeatin. 2iP has been used in Schenk and Hildebrandt medium to support in vitro propagation of microshoot cultures from shoot tips of Genista plants.
6-(γ,γ-Dimethylallylamino)purine has been used as a component of Murashige and Skoog medium for promoting tissue lignification in Acca sellowiana and as a component of SIM medium for used in regeneration assay of cotyledons.
Biochem/physiol Actions
6-(γ,γ-Dimethylallylamino)purine (2iP) is a bacteria-derived riboside cytokinin used to grow plant tissues such as tobacco and soybean callus. 2iP is a precursor of the cytokinin Zeatin. 2iP is used in Schenk and Hildebrandt medium to support in vitro propagation of microshoot cultures from shoot tips of Genista plants.
Still not finding the right product?
Explore all of our products under 6-(γ,γ-Dimethylallylamino)purine
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The shoot regeneration capacity of excised Arabidopsis cotyledons is established during the initial hours after injury and is modulated by a complex genetic network of light signalling
Nameth B, et al.
Plant, Cell and Environment, 36(1), 68-86 (2013)
In vitro propagation of Guayabo del pais(Acca sellowiana (Berg.) Burret)
Ross S and Grasso R
Fruit, Vegetable and Cereal Science and Biotechnology, 4, 83-87 (2010)
6-gamma, gamma-Dimethylallylamino) purine as a precursor of zeatin
Miura GA and Miller CO
Plant Physiology, 44(3), 372-376 (1969)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| D7674-1G | 04061833588727 |
| D7674-5G | 04061833588734 |
| D7674-10G | 04061833588710 |