Skip to Content
Merck
All Photos(6)

Key Documents

B3408

Sigma-Aldrich

6-Benzylaminopurine

suitable for plant cell culture

Synonym(s):

6-BAP, BA, N6-Benzyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5
CAS Number:
Molecular Weight:
225.25
Beilstein:
19406
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

form

powder

Quality Level

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

C(Nc1ncnc2nc[nH]c12)c3ccccc3

InChI

1S/C12H11N5/c1-2-4-9(5-3-1)6-13-11-10-12(15-7-14-10)17-8-16-11/h1-5,7-8H,6H2,(H2,13,14,15,16,17)

InChI key

NWBJYWHLCVSVIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-Benzylaminopurine is a plant growth regulator that belongs to the class of first generation synthetic cytokinin used in agriculture.

Application

6-Benzylaminopurine has been used:
  • to induce sprouting in plant materials
  • in seed germination medium for culturing of seeds
  • to modify Murashige and Skoog (MS) media for shoot initiation

6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin which together with auxins elicits plant growth and development responses. BAP is a widely use cytokinin supplement to plant growth media such as Murashige and Skoog medium, Gamborg′s medium, and Chu′s N6 medium.

Biochem/physiol Actions

BAP is an inhibitor of respiratory kinase in plants, and increases post-harvest life of green vegetables.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro plant regeneration from commercial cultivars of soybean
Raza G, et al.
BioMed Research International, 2017 (2017)
Callus induction and organogenesis in soybean [Glycine max (L.) Merr.] cv. Pyramid from mature cotyledons and embryos
Joyner E Y, et al.
The Open Plant Science Journal, 4(1) (2010)
In vitro propagation of `Guayabo del pais?(Acca sellowiana (Berg.) Burret)
Ross S and Grasso R
Fruit, Vegetable and Cereal Science and Biotechnology, 4(special issue 1), 83-87 (2010)
Sophie Donovan et al.
Frontiers in genome editing, 2, 605614-605614 (2021-10-30)
Engineering the small subunit of the key CO2-fixing enzyme Rubisco (SSU, encoded by rbcS) in plants currently poses a significant challenge, as many plants have polyploid genomes and SSUs are encoded by large multigene families. Here, we used CRISPR-Cas9-mediated genome
Monica Carabelli et al.
Nature communications, 12(1), 4321-4321 (2021-07-16)
Symmetry establishment is a critical process in the development of multicellular organs and requires careful coordination of polarity axes while cells actively divide within tissues. Formation of the apical style in the Arabidopsis gynoecium involves a bilateral-to-radial symmetry transition, a

Protocols

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service