Skip to Content
Merck
All Photos(5)

Key Documents

T9128

Sigma-Aldrich

Trypsin inhibitor

lyophilized powder, suitable for inhibition assay

Synonym(s):

SBTI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Trypsin inhibitor from Glycine max (soybean), lyophilized powder

biological source

Glycine max (soybean)

Quality Level

type

Type II-S

form

lyophilized powder

mol wt

20,100 Da

technique(s)

inhibition assay: suitable

solubility

balanced salt solution: 1 mg/mL
concentrate: >10 mg/mL, hazy, amber-yellow
phosphate buffer: 10 mg/mL
water: 10 mg/mL
serum-free medium: soluble

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

This inhibitor acts against trypsin, and chymotrypsin and plasmin to a lesser extent. It will also inhibit proteases with mechanisms similar to trypsin, plasma kallikrein and coagulation Factor X. The trypsin inhibitor will not act against metalloproteases, tissue-baseed kallikrein, acid proteases, or thio proteases. This inhibitor acts by forming a 1:1 stoichiometric complex with the protease active site, and then cleaving a single arginine-isoleucine bond on the inhibitor. The inhibition is both reversible and pH dependent.

Components

The soybean trypsin inhibitor is a monomeric protein containing 181 amino acid residues in a single polypeptide chain crosslinked by two disulfide bridges.

Unit Definition

One trypsin unit = A253 of 0.001 per minute with N-alpha-benzoyl-L-arginine ethyl ester (BAEE) as substrate at pH 7.6 at 25 °C.
One trypsin unit will produce a ΔA253 of 0.001 per min with BAEE as substrate at pH 7.6 at 25 °C; reaction volume 3.2 ml, 1 cm light path.

Preparation Note

The trypsin inhibitor is soluble in water and phosphate buffers at 10 mg/mL. It is soluble in balanced salt solutions at 1 mg/mL and in serum-free media. Concentrated solutions greater than 10 mg/mL may be hazy and have a yellow to amber color. After trypsinizing cells, resuspend in 1 mL trypsin inhibitor solution at 1 mg/mL for every mL of trypsin solution used for dissociation. The cell suspension should then be centrifuged at 1000 rpm, forming a cell pellet.

Solutions can retain activity when stored short-term at 2-8° C. Solutions are stable in frozen aliquots at -20°C.

Analysis Note

One mg of trypsin inhibitor will inhibit a minimum of 1.0 mg Trypsin of activity ~10,000 BAEE units/mg trypsin.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dolores D Mruk et al.
Methods in molecular biology (Clifton, N.J.), 763, 237-252 (2011-08-30)
The use of an in vitro system based on primary cultures of Sertoli cells isolated from rat testes has greatly facilitated the study of the blood-testis barrier in recent years. Herein, we summarize the detailed procedures on the isolation of
I T Padol et al.
Infection and immunity, 69(6), 3891-3896 (2001-05-12)
As a bacterial product, Helicobacter pylori lipopolysaccharide (LPS) can originate in close proximity to parietal cells, but the role of this uniquely structured endotoxin on acid secretion has not been fully investigated and remains unclear. The purpose of this study
Ho Am Jang et al.
Developmental and comparative immunology, 67, 97-106 (2016-11-09)
Riptortus pedestris, a common pest in soybean fields, harbors a symbiont Burkholderia in a specialized posterior midgut region of insects. Every generation of second nymphs acquires new Burkholderia cells from the environment. We compared in vitro cultured Burkholderia with newly in vivo
Patrik Hollos et al.
eNeuro, 7(1) (2020-01-16)
In this study, we use an optogenetic inhibitor of c-Jun NH2-terminal kinase (JNK) in dendritic spine sub-compartments of rat hippocampal neurons. We show that JNK inhibition exerts rapid (within seconds) reorganization of actin in the spine-head. Using real-time Förster resonance
Anne-Rose Denet et al.
Journal of controlled release : official journal of the Controlled Release Society, 88(2), 253-262 (2003-03-12)
The purpose was to achieve therapeutic fluxes of timolol by transdermal delivery using skin electroporation. The transdermal transport of timolol through human stratum corneum was studied in three compartment diffusion cells. The electrodes, buffer composition and pulse conditions were optimized.

Protocols

This technical article described the Enzymatic Assay of Trypsin Inhibitor.

Chromatograms

application for HPLC

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the molecular weight and amino acid sequence of trypsin inhibitor from Glycine max (soybean)?

    1 answer
    1. Soybean trypsin inhibitor is a mixture of the three types. We have not characterized our trypsin inhibitor products in terms of how much of each of the three types may be present. All three have a molecular weight of between 20,000 and 20,200 daltons, so one might use 20,100 daltons as an average.The three types vary in the amino acids at a few positions. The detailed summary of the three types (or variants) was published in the following linked reference:     J. Biochem. (Japan), 98, 435-448 (1985).Note that the authors use Tia, Tib and Tic to indicate the three variants.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service