Skip to Content
Merck
All Photos(1)

Key Documents

10109886001

Roche

Trypsin Inhibitor

from soybean

Synonym(s):

trypsin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

soybean

Quality Level

form

lyophilized

mol wt

Mr 20,100

packaging

pkg of 50 mg

manufacturer/tradename

Roche

concentration

1 mg/mL

technique(s)

cell culture | mammalian: suitable

pH

3.8-4.5

solubility

water: soluble

storage temp.

2-8°C

General description

Soybean trypsin inhibitor (STI) comprises 181 amino acids. It is a monomeric protein corresponding to a molecular weight of 20 kDa and has two disulfide bridges. It belongs to serpins − serine protease inhibitors family.

Specificity

Soybean trypsin inhibitor is an inhibitor for trypsin, plasmin, and plasma kallikrein. It inhibits trypsin, factor Xa, plasmin, and plasma kallikrein activity in serum-free cell culture media. It does not inhibit metallo-, cysteine, aspartic proteases, or tissue kallikrein (serine proteases).

Application

Trypsin Inhibitor from soybean is used to terminate tissue disaggregation and for subcultivation procedures.
Trypsin Inhibitor has been used:
  • in the buffer for hepatocyte isolation
  • to inhibit trypsin in Corneal epithelial cells
  • to inhibit papain activity while treating hippocampal neural stem cells (NSCs)

Biochem/physiol Actions

Soybean trypsin inhibitor (STI) inhibits myc proto-oncogene protein and is anticarcinogenic. It favors anti-aggregation of platelets and may be a potential protease inhibiting drug. STI inhibits renin and angiotensin.
Soybean trypsin inhibitor is an inhibitor for trypsin, plasmin, and plasma kallikrein. The product is sensitive to heat, alkaline pH, and protein-precipitating compounds.

Specifications

Inhibiting activity: approximately 70 trypsin inhibitor units/mg lyophilizate at +25°C with benzoyl-L-arginine ethylester as a substrate (approximately 190 trypsin inhibitor units/mg at +25°C with Chromozym TRY as substrate)

Isoelectric point: 4.5

Unit Definition

1 U = 270 S & T-units or 90 USP-units

Preparation Note

Working concentration: 10 to 100 μg/ml
Working solution: Recommended solvent is distilled water.
Stock solution: 1 mg/ml
Storage conditions (working solution): -15 to -25 °C
The enzyme is stable for 6 months, frozen in aliquots.

Reconstitution

Sensitive to heat, alkaline pH, and protein-precipitating compounds.

Storage and Stability

Store at 2 to 8 °C. (Store dry!)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolating Taste Buds and Taste Cells from Vallate Papillae of C57BL/6J Mice for Detecting Transmitter Secretion
Anthony Y. Huang and Sandy Y. Wu
CorpBase ID (for auto-filling citation data), 6(11) (2016)
Benjamin C Orsburn et al.
Nature communications, 13(1), 7246-7246 (2022-11-27)
Single cell proteomics is a powerful tool with potential for markedly enhancing understanding of cellular processes. Here we report the development and application of multiplexed single cell proteomics using trapped ion mobility time-of-flight mass spectrometry. When employing a carrier channel
Preparation of Primary Rat Hepatocyte Spheroids Utilizing the Liquid-Overlay Technique
Kyffin JA, et al.
Current Protocols in Toxicology / Editorial Board, Mahin D. Maines (editor-in-chief) ... [Et al.], 81(1), e87-e87 (2019)
Single cell transcriptomic analysis of renal allograft rejection reveals insights into intragraft TCR clonality.
Shi, et al.
The Journal of Clinical Investigation (2023)
Veronica Kon Graversen et al.
Methods in molecular biology (Clifton, N.J.), 1357, 395-401 (2014-11-19)
Reprogramming fibroblasts into induced pluripotent stem cells (iPSC) remains a promising technique for cell replacement therapy. Diverse populations of somatic cells have been examined for their reprogramming potential. Recently, ocular ciliary body epithelial cells (CECs) have been reprogrammed with high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service