Skip to Content
Merck
All Photos(1)

Key Documents

T7659

Sigma-Aldrich

Trypsin Inhibitor, Defined (1X) Solution

Animal component free, BioReagent, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75
Pricing and availability is not currently available.

sterility

sterile-filtered

product line

BioReagent

form

solution

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

Application

This product designed to be used during the subculture of keratinocytes contains Kunitz-type soybean trypsin inhibitor (SBTI) which inhibits the catalytic activity of serine proteases such as trypsin and tryptase. It is used in cell culture to stop the action of trypsin which is used to release cells from substratum during passaging. It may be also useful in other anti-serine protease studies.
Used to neutralize the effects of trypsin/EDTA solution.

Physical form

Solution in 98.7% DPBS and 1.3% Soybean Trypsin Inhibitor.

Reconstitution

Use at a minimum volume ratio of 1:1 TID:trypsin/EDTA.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarra Djemil et al.
Cellular and molecular neurobiology, 41(8), 1787-1799 (2020-08-30)
Septal innervation of basal forebrain cholinergic neurons to the hippocampus is critical for normal learning and memory and is severely degenerated in Alzheimer's disease. To understand the molecular events underlying physiological cholinergic synaptogenesis and remodeling, as well as pathological loss
Maurizio Trovato et al.
Biochemical and biophysical research communications, 302(2), 311-315 (2003-02-27)
The design of minimal units required for enzyme inhibition is a major field of interest in structural biology and biotechnology. The successful design of the cyclic dodecapeptide corresponding to the Phe17-Val28 reactive site amino acid sequence of the low-molecular-mass trypsin
I B Svendsen et al.
Carlsberg research communications, 54(6), 231-239 (1989-01-01)
A trypsin inhibitor with a Km of 5 x 10(-5) M has been isolated from kohlrabi (Brassica napus var. rapifera). Subtilisin DY is inhibited only weakly and chymotrypsin not at all. The inhibitor is closely related to napin as determined
C L Dumke et al.
Journal of applied physiology (Bethesda, Md. : 1985), 92(2), 657-664 (2002-01-18)
Serum proteins [molecular weight (MW) > 10,000] are essential for increased insulin-stimulated glucose transport after in vitro muscle contractions. We investigated the role of the kallikrein-kininogen system, including bradykinin, which is derived from kallikrein (MW > 10,000)-catalyzed degradation of serum

Protocols

Natural trypsin inhibitors (serpins) regulate protein activation and catabolism by inhibiting serine proteases in vivo.

Natural trypsin inhibitors (serpins) regulate protein activation and catabolism by inhibiting serine proteases in vivo.

Natural trypsin inhibitors (serpins) regulate protein activation and catabolism by inhibiting serine proteases in vivo.

Natural trypsin inhibitors (serpins) regulate protein activation and catabolism by inhibiting serine proteases in vivo.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service