Skip to Content
Merck
All Photos(1)

Documents

T0637

Sigma-Aldrich

Trypsin inhibitor

saline suspension

Synonym(s):

Trypsin Inhibitor Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

product name

Trypsin inhibitor–Agarose, saline suspension, protein from Glycine max (soybean)

biological source

protein from Glycine max (soybean)

form

saline suspension

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥1 mg/mL binding capacity (trypsin)(with activity of 10,000 BAEE units per mg)

storage temp.

2-8°C

Application

Trypsin inhibitor-Agarose has been used in affinity chromatography for the purification:

  • of shrimp chymotrypsin
  • of protease from Trichoderma reesei
  • of Ras-interacting protein 1(Rasip 1)
Trypsin inhibitor-agarose is used in protein chromatography, affinity chromatography, and specialty resins.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D P Goldenberg et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(11), 5083-5087 (1992-06-01)
In a previous study, a genetic screening procedure was used to identify variants of bovine pancreatic trypsin inhibitor that can fold to an active conformation but that are inactivated much more rapidly than the wild-type protein in the presence of
J S Munger et al.
Molecular biology of the cell, 9(9), 2627-2638 (1998-09-03)
The multipotential cytokine transforming growth factor-beta (TGF-beta) is secreted in a latent form. Latency results from the noncovalent association of TGF-beta with its processed propeptide dimer, called the latency-associated peptide (LAP); the complex of the two proteins is termed the
Shuishu Wang et al.
Protein science : a publication of the Protein Society, 12(5), 1097-1108 (2003-04-30)
Pantothenate biosynthesis is essential for the virulence of Mycobacterium tuberculosis, and this pathway thus presents potential drug targets against tuberculosis. We determined the crystal structure of pantothenate synthetase (PS) from M. tuberculosis, and its complexes with AMPCPP, pantoate, and a
D Lu et al.
Journal of molecular biology, 292(2), 361-373 (1999-09-24)
Enteropeptidase is a membrane-bound serine protease that initiates the activation of pancreatic hydrolases by cleaving and activating trypsinogen. The enzyme is remarkably specific and cleaves after lysine residues of peptidyl substrates that resemble trypsinogen activation peptides such as Val-(Asp)4-Lys. To
S Lawler et al.
Current biology : CB, 8(25), 1387-1390 (1999-01-16)
Mitogen-activated protein kinases (MAPKs) mediate many of the cellular effects of growth factors, cytokines and stress stimuli. Their activation requires the phosphorylation of a threonine and a tyrosine residue located in a Thr-X-Tyr motif (where X is any amino acid)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service