Skip to Content
Merck
All Photos(1)

Key Documents

SRP3326

Sigma-Aldrich

BMP-2 human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

BMP-2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

26.0 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BMP2(650)

General description

The functional form of BMP-2 (bone morphogenetic protein 2) is a 26kDa protein composed of two identical 114 amino-acid polypeptide chains (monomers) linked by a single disulfide bond. Each BMP-2 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 23 amino-acid signal sequence for secretion, and a 259 amino-acid propeptide. After dimerization of this precursor, the covalent bonds between the propeptide (which is also a disulfide-linked homodimer) and the mature BMP-2 ligand are cleaved by a furin-type protease. Recombinant human BMP-2 is a 26kDa homodimeric disulfide-linked protein consisting of two identical 115 amino acid chains.

Application

BMP-2 human has been used to study the biological role of BMP-2 on adult renal progenitor cells under in vitro and in vivo conditions.

Biochem/physiol Actions

BMP-2 (bone morphogenetic protein 2) is a potent osteoinductive cytokine, capable of inducing bone and cartilage formation in association with an osteoconductive carrier such as collagen and synthetic hydroxyapatite. During bone repair, it controls the expression of Runt-related transcription factor 2 (Runx2) and Osterix (Osx), thereby enhancing migration, proliferation and osteogenic differentiation of mesenchymal stem cells. In addition to its osteogenic activity, BMP-2 appears to play an important role in cardiac morphogenesis, and is expressed in a variety of other tissues including lung, liver, spleen, prostate, ovary, and small intestine.

Physical form

Lyophilized from 0.1% TFA.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bone regeneration with low dose BMP-2 amplified by biomimetic supramolecular nanofibers within collagen scaffolds.
Lee SS, et al.
Biomaterials, 34, 452-459 (2013)
Bone morphogenetic proteins: multifunctional regulators of vertebrate development.
Hogan BL
Genes & Development, 10, 1580-1594 (1996)
A role for bone morphogenetic proteins in the induction of cardiac myogenesis.
Schultheiss TM, et al.
Genes & Development, 11, 451-462 (1997)
Vera Hintze et al.
Biomacromolecules, 15(8), 3083-3092 (2014-07-17)
Sulfated glycosaminoglycans (GAGs) can direct cellular processes by interacting with proteins of the extracellular matrix (ECM). In this study we characterize the interaction profiles of chemically sulfated hyaluronan (HA) and chondroitin sulfate (CS) derivatives with bone morphogenetic protein-2 (BMP-2) and
Rick Visser et al.
Tissue engineering. Part A, 20(1-2), 34-44 (2013-07-19)
Osteogenesis is a complex, multifactorial process in which many different signals interact. The bone morphogenetic proteins (BMPs) are the most potent inducers of osteoblastic differentiation, although very high doses of BMPs in combination with collagen type I formulations have to

Protocols

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service