Skip to Content
Merck
All Photos(1)

Documents

HT501128

Sigma-Aldrich

Formalin solution, neutral buffered, 10%

histological tissue fixative

Synonym(s):

Formaldehyde, Methanal

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.47

biological source

synthetic

concentration

10% formalin (approx. 4% formaldehyde)

color

colorless

pH

6.90-7.10

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

10% Neutral buffered formalin (NBF) is the most commonly used fixative throughout the world for light microscopy and is a somewhat forgiving fixative. 10% NBF is well suited for large throughput laboratories, and requires a relatively short period of fixation, but can also be used for the long-term storage of tissue. Formalin fixation is thought to form between the aldehydes and the proteins, creating a gel, thus retaining cellular constituents in their in vivo relationship. Once properly fixed, the tissue should be able to withstand the subsequent stages of tissue processing or staining.

Application

Formalin solution, neutral buffered, 10% has been used as a popular general purpose histological fixative to fix tissues.
Formalin solution, neutral buffered, 10% has been used in oil-red-o staining for the differentiation of 3T3-L1 preadipocyte cell line and mouse embryonic fibroblasts into adipocytes. It has been used in the histological analysis of rabbit′s eye and diabetic rat intestine.
Popular general purpose histological fixative.

Physical form

Neutral buffered solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Recapitulating spatiotemporal tumor heterogeneity in vitro through engineered breast cancer microtissues.
Mazio c, et al.
Acta Biomaterialia, 73, 236-249 (2018)
Comparison of partially and fully chemically-modified siRNA in conjugate-mediated delivery in vivo.
Hassler M R, et al.
Nucleic Acids Research, 46(5), 2185-2196 (2018)
Paul J Marchand et al.
Biomedical optics express, 8(7), 3343-3359 (2017-07-19)
We present a novel extended-focus optical coherence microscope (OCM) attaining 0.7 μm axial and 0.4 μm lateral resolution maintained over a depth of 40 μm, while preserving the advantages of Fourier domain OCM. Our system uses an ultra-broad spectrum from
Ablation of the transcriptional regulator Id1 enhances energy expenditure, increases insulin sensitivity, and protects against age and diet induced insulin resistance, and hepatosteatosis
Satyanarayana A, et al.
Faseb Journal, 26(1), 309-323 (2012)
Effects of the marine microalgae Isochrysis galbana and Nannochloropsis oculata in diabetic rats
Nu?o K, et al.
Journal of functional foods, 5(1) (2013)

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service