Skip to Content
Merck
All Photos(1)

Key Documents

T7039

Sigma-Aldrich

Transforming Growth Factor-β1 human

≥98% (SDS-PAGE), recombinant, expressed in CHO cells, powder, suitable for cell culture

Synonym(s):

hTGF-β1, TGF-β1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in CHO cells

Assay

≥98% (SDS-PAGE)

form

powder

potency

≤0.05 ng/mL ED50

quality

endotoxin tested

mol wt

protein 25 kDa

packaging

pkg of 2 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TGFB1(7040)

Looking for similar products? Visit Product Comparison Guide

General description

Tgfb1 (transforming growth factor-β1 human) is a 25kDa peptide encoded by the gene mapped to human chromosome 19q13.2.[1][2] TGF-β1 polypeptide is produced as a precursor with hydrophobic signal sequence, pro-region and mature peptide. Cleavage of the precursor at tetrabasic cleavage site results in monomers of carboxy-terminal 112 amino acids. The biologically active form of Tgfb1 exists as a disulphide-linked homodimer. In mammals, three isoforms of TGF-β has been identified : TGF-β1, TGF-β2 and TGF-β3.[3]

Application

Transforming Growth Factor-β1 human has been used in
  • reporter gene assay.[4]
  • in vitro transfection.[5]
  • to study the effect of transforming growth factor-β1 (TGF-β1) on gene array analysis of renal cells.[6]
  • to study the association of TGF-β1 with shear stress-dependent upregulation of tissue inhibitor of metalloproteinase-1 in microvascular endothelial cells.[7]
  • to study the interaction between TGF-β1 activation and myofibroblast formation.[8]

Biochem/physiol Actions

TGF-β1 is a multifunctional peptide capable of influencing cell proliferation, tissue growth, differentiation, and other functions in a wide range of cell types. TGF-β1 plays a fundamental role in differentiation by involvement in adipogenesis, myogenesis, chondrogenesis, osteogenesis, epithelial cell differentiation and immune cell function.[9] All cells possess a specific TGF-β1 receptor. Both transformed and non-neoplastic tissues release transforming growth factors.The multi-modal nature of TGF-β1 is seen in its ability to stimulate or inhibit cellular proliferation.[10] TGF-β1 is associated with the developmental and metastasis of colorectal cancer.[11]
Transforming growth factor-β1 (TGF-β1) is produced by many cell types, but is reported to be most concentrated in mammalian platelets, where it is present at approximately four times the level of TGF-β2.

Physical form

Lyophilized from a 0.2 μm filtered aqueous solution.

Analysis Note

The biological activity of TGF-β1 is measured in culture by inhibition of mouse IL-4-dependent proliferation of mouse HT-2 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix metalloproteinase inhibition delays wound healing and blocks latent transforming growth factor-β1 promoted myofibroblast formation and function
Mirastschijski U, et al.
Wound Repair and Regeneration, 18(2), 223-234 (2010)
Interplay between TGF-β signaling and receptor tyrosine kinases in tumor development.
Shi Q and Chen YG
Science China: Life Sciences, 60(10), 1133-1141 (2017)
Tibor Kempf et al.
Clinical chemistry, 53(2), 284-291 (2006-12-23)
Growth-differentiation factor 15 (GDF15) is a member of the transforming growth factor beta (TGF-beta) cytokine superfamily. There has been increasing interest in using circulating GDF15 as a biomarker in patients, for example those with cardiovascular disease. We developed an IRMA
TGF β signalling and its role in tumour pathogenesis.
Kaminska B, et al.
Acta Biochimica Polonica, 52(2), 329-337 (2005)
Louis A Saddic et al.
Cardiovascular diabetology, 16(1), 87-87 (2017-07-09)
Omentin-1, also known as Intelectin-1 (ITLN1), is an adipokine with plasma levels associated with diabetes, obesity, and coronary artery disease. Recent studies suggest that ITLN1 can mitigate myocardial ischemic injury but the expression of ITLN1 in the heart itself has

Articles

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

Naive pluripotent stem cells cultured in vitro using specialized media and inhibitors mimic "ground-state" cells from blastocysts.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service