Skip to Content
Merck

GF166

BMP-2 Protein, Human Recombinant

The Human Bone morphogenetic protein 2 (or BMP-2 protein) is a member of the BMP protein subgroup of the TGFβ superfamily.

Synonym(s):

Bone Morphogenetic Protein 2, BMP2, BMP-2A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405
Assay:
>98% (SDS-PAGE and HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Bone morphogenetic protein 2 (BMP-2) is a member of the BMP subgroup of the TGFβ superfamily. It plays a dominant role in embryonic dorsalventral patterning, organogenesis, limb bud formation, and bone formation and regeneration. BMP-2 is a potent osteoinductive cytokine, capable of inducing bone and cartilage formation in association with osteoconductive carriers such as collagen and synthetic hydroxyapatite. In addition to its osteogenic activity, BMP-2 plays an important role in cardiac morphogenesis and is expressed in a variety of tissues including lung, spleen, brain, liver, prostate ovary and small intestine. The functional form of BMP-2 is a 26 kDa protein composed of two identical 114 amino acid polypeptide chains linked by a single disulfide bond. Each BMP-2 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 23 amino acid signal sequence for secretion, and a 259 amino acid propeptide. After dimerization of this precursor, the covalent bonds between the propeptide (which is also a disulfide-linked homodimer) and the mature BMP-2 ligand are cleaved by a furin-type protease. Recombinant human BMP-2 is a 26.0 kDa homodimeric protein consisting of two 115 amino acid polypeptide chains.
Product Source: E. coli.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Biochem/physiol Actions

Cross Reactivty
Mouse, Rat, Rabbit, Cow, Dog, Monkey

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.

Preparation Note

Store at -20°C for up to 6 months from date of receipt.

Analysis Note

Bioactivity assay: Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.0 μg/ml.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins


Xingtao Ge et al.
American journal of translational research, 13(6), 6330-6341 (2021-07-27)
To investigate the protective effects of Naringin on glucocorticoid-induced osteoporosis (GIOP) through the PI3K/AKT/mTOR signaling pathway in vivo and in vitro. Osteoblasts were cultured from the differentiated bone marrow mesenchymal stem cells (BM-MSCs) and were grouped as follows: the PBS



Global Trade Item Number

SKUGTIN
GF16604053252691232