Skip to Content
Merck
All Photos(2)

Documents

SAB5200111

Sigma-Aldrich

Anti-AQP3 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-AQP3, Anti-Aquaporin 3, Anti-Aquaporin 3 (GIL blood group), Anti-GIL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 31.5 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Aqp3(65133)

General description

The gene ID of the protein aquaporin-3 (AQP3) is 65133.

Specificity

Detects ~31.5 kDa. May detect larger glycosylated bands ~35-50 kDa.

Immunogen

C-terminal peptide

Biochem/physiol Actions

Aquaporin 3 (AQP3) deals with transportation of water and cryoprotectants in rats.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, pH 7..4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Farrell et al.
Physiological reports, 7(24), e14314-e14314 (2019-12-25)
Facilitative UT-B urea transporters play important physiological roles in numerous tissues, including the urino-genital tract. Previous studies have shown that urothelial UT-B transporters are crucial to bladder function in a variety of mammalian species. Using the RT4 bladder urothelial cell
Giorgia Pellavio et al.
Cells, 9(5) (2020-05-21)
Human sperm cells express different aquaporins (AQPs), AQP3, 7, 8, 11, which are localized both in the plasma membrane and in intracellular structures. Besides cell volume regulation and end stage of cytoplasm removal during sperm maturation, the role of AQPs
Chongliang Zhong et al.
Journal of dairy science, 103(3), 2814-2820 (2020-01-26)
Urea nitrogen salvaging is a crucial mechanism that ruminants have evolved to conserve nitrogen. Facilitative urea transporter-B proteins are known to be involved in urea transport across the rumen epithelium and thus efficiently facilitate the urea nitrogen salvaging process. Recently
Chongliang Zhong et al.
Veterinary sciences, 9(2) (2022-02-25)
Rumen studies have focused almost exclusively on livestock species under strictly regimented diets. This means that the ruminal condition of free-living and free-feeding wildlife remains practically unstudied. Urea nitrogen salvaging, a process by which urea is passed into the rumen
Simona Martinotti et al.
Life (Basel, Switzerland), 10(11) (2020-10-31)
Honey is a natural product with a long use in traditional medicine and is well recognized to regulate different biological events. It is an important source of various biological or pharmacological molecules and, therefore, there is a strong interest to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service