Skip to Content
Merck

A0303

Anti-Water Channel Aquaporin 3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

anti-AQP3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable using Mouse kidney sections, western blot: 1:200 using Rat kidney membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Aqp3(11828)
rat ... Aqp3(65133)

General description

Aquaporins function in the movement of water and small solutes across the cell membrane. Aquaporins are widely distributed from bacteria to plants and animals. In E. coli, one aquaporin has been identified that is named AQP-Z. In the plant Arabidopsis thaliana, over 9 AQPs have been characterized. In mammals, at least 11 different AQPs have been cloned. AQP3 provides kidney medullary collecting ducts with high permeability to water, permitting water to move in the direction of an osmotic gradient and may play an important role in gastrointestinal tract water transport. AQP3 and AQP4 are located at the basolateral part of epithelial cells in kidney, colon and trachea.
Rabbit polyclonal anti-Water Channel Aquaporin 3 recognizes rat and mouse aquaporin 3 transport proteins.

Immunogen

synthetic peptide corresponding to amino acids 275-292 of rat aquaporin 3 (with additional N-terminal lysine and tyrosine) conjugated to KLH with glutaraldehyde

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Rabbit anti-AQP3 antibody was used in western blot analysis to identify the level of aquaporin 3 in E. solidaginis larvae. Anti-AQP3 was diluted 1:200 in 5% nonfat dry milk and incubated with the membrane for 2hrs at 21 degrees C.
Rabbit polyclonal anti-Water Channel Aquaporin 3 is used to tag aquaporin 3 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of aquaporin 3 in kidney medullary collecting ducts and the basolateral surfaces of epithelial cells involved in water transport.

Physical form

Lyophilized from a solution containing phosphate buffered saline, pH 7.4, 1% BSA, and 0.05% sodium azide

Preparation Note

Affinity isolated on immobilized Protein A

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library