Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500813

Sigma-Aldrich

Anti-Estrogen Receptor-α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ER, ESR, ESR1, ESTR, ESTRA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 66 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

General description

Anti-Estrogen Receptor-α Antibody detects endogenous levels of total Estrogen Receptor-α protein.
Estrogen receptor α (ERα) is a member of the nuclear receptor superfamily. It is also known as ESR1 (estrogen receptor). It is located on human chromosome 6q25.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Estrogen Receptor-alpha.

Immunogen Range: 136-185

Biochem/physiol Actions

Estrogen receptor α (ERα) is involved in immune and inflammatory response. Aberrations in ESR1 results in Mayer-Rokitansky-Küster-Hauser syndrome that is associated with the congenital absence of the uterus and the upper two-thirds of the vagina.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

ERα36, a variant of estrogen receptor α, is predominantly localized in mitochondria of human uterine smooth muscle and leiomyoma cells
Yan Y, et al.
PLoS ONE, 12(10), e0186078-e0186078 (2017)
Amplification of WHSC1L1 regulates expression and estrogen-independent activation of ER? in SUM-44 breast cancer cells and is associated with ER? over-expression in breast cancer
Irish JC, et al.
Molecular Oncology, 10(6), 850-865 (2016)
Anita K Dunbier et al.
PLoS genetics, 7(4), e1001382-e1001382 (2011-05-10)
Approximately 80% of human breast carcinomas present as oestrogen receptor α-positive (ER+ve) disease, and ER status is a critical factor in treatment decision-making. Recently, single nucleotide polymorphisms (SNPs) in the region immediately upstream of the ER gene (ESR1) on 6q25.1
Sequence variants in ESR1 and OXTR are associated with Mayer-Rokitansky-Kuster-Hauser syndrome
Brucker SY, et al.
Acta Obstetricia et Gynecologica Scandinavica, 96(11), 1338-1346 (2017)
Jun Wang et al.
Discover. Oncology, 14(1), 28-28 (2023-03-01)
Circumvention of apoptosis by the elevation of antiapoptotic proteins is an important cause of carcinogenesis. The induction of antiapoptotic genes, including B-cell CLL/lymphoma 2 (BCL2), BCL2 related protein A1 (BCL2A1), BCL2 like 1 (BCL2L1), BCL2L2, and myeloid cell leukemia 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service