Skip to Content
Merck
All Photos(1)

Key Documents

B5645

Sigma-Aldrich

Anti-Bovine IgG (whole molecule) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:20,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG (immunoglobulin G) contributes to 10-20% of plasma protein and is regarded as one of the most predominant serum proteins. It consists of four subclasses: IgG1, IgG2, IgG3 and IgG4. The IgG structure possesses four polypeptide chains containing two identical γ heavy (H) chains and two identical κ or λ light (L) chains of 50 kDa and 25 kDa, respectively.
Immunoglobulin G (IgG) is the most important type of glycoprotein antibody present in human serum. It is found in blood and has four polypeptide chains.The light and heavy chains are interconnected by disulfide bonds. IgG has the longest serum half-life when compared to other immunoglobulins.

Application

Anti-Bovine IgG (whole molecule) antibody produced in rabbit has been used to coat immulon 4HBX plates in serum antibody enzyme-linked immunosorbent assay (ELISA) and western blotting.
Anti-Bovine IgG (whole molecule) antibody produced in rabbit is suitable for ELISA at a working dilution of 1:20,000. It may be used for immunohistochemistry of formalin-fixed, paraffin-embedded sections at a dilution of 1:1000.

Biochem/physiol Actions

Immunoglobulin G (IgG) initiates the classical pathway of the complement system and also nullifies virus particles and toxins. Maternal IgG is transferred to the fetus through the placenta that is critical for the immune defense of the neonate against infections. IgG modulates antibody-dependent cell-mediated cytotoxicity (ADCC) and might be associated with allergy. Digestion of IgG by papain releases fragment antigen-binding (Fab) and pepsin digestion of IgG produces crystallizable (fc) fragment and a single dimeric F(ab)2. IgG levels in severe acute respiratory syndrome coronavirus (SARS-CoV)-2 infected patients may be used to evaluate the severity and prognosis of coronavirus disease 2019 (COVID-19).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Spiesberger et al.
PloS one, 10(10), e0140471-e0140471 (2015-10-16)
30 years ago, the potential of bispecific antibodies to engage cytotoxic T cells for the lysis of cancer cells was discovered. Today a variety of bispecific antibodies against diverse cell surface structures have been developed, the majority of them produced
Low expression of secreted frizzled-related protein 2 and nuclear accumulation of beta-catenin in aggressive nonfunctioning pituitary adenoma
Wu Y, et al.
Oncology Letters, 12(1), 199-206 (2016)
B W Blais
Journal of food protection, 62(4), 386-389 (1999-07-27)
An enzyme immunoassay system based on the use of a macroporous swab as a solid phase for the capture and subsequent immunoenzymatic detection of immunoglobulin G (IgG) from meat residues on food contact surfaces was developed as a hygiene-monitoring tool.
Formalin-inactivated bovine RSV vaccine enhances a Th2 mediated immune response in infected cattle
Kalina WV, et al.
Vaccine, 22(11-12), 1465-1474 (2004)
Transfer of maternal colostral leukocytes promotes development of the neonatal immune system: Part II. Effects on neonatal lymphocytes
Reber AJ, et al.
Veterinary Immunology and Immunopathology, 123(3-4), 305-313 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service