Skip to Content
Merck
All Photos(3)

Documents

09569

Supelco

Span® 80

greener alternative

suitable for GC

Synonym(s):

Sorbitan monooleate, Span® 80

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H44O6
CAS Number:
Molecular Weight:
428.60
Beilstein:
8172514
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

mol wt

428.60 g/mol

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

LPLC: suitable
gas chromatography (GC): suitable
protein quantification: suitable

refractive index

n20/D 1.48 (lit.)

HLB value

4.6±0.1

density

0.99 g/mL at 20 °C
0.986 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[H][C@@](O)(COC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@H](O)[C@H]1O

InChI

1S/C24H44O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h9-10,20-21,23-26,28H,2-8,11-19H2,1H3/b10-9-/t20-,21+,23+,24+/m0/s1

InChI key

NWGKJDSIEKMTRX-AAZCQSIUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is biodegradable surfactant, thus aligns with "Use of Renewable Feedstocks" and "Design for Degradation" principles of Green Chemistry, and belongs to "12 principles aligned" category of our greener alternatives.

Application

A non-ionic surfactant that can be used to form oil-in-water emulsions and in combination with low HLB surfactants can form water-in-oil emulsions. EPA approved oil spill dispersant.

Legal Information

Span is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hoang Hirschberg et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 46(1-2), 1-7 (2012-02-15)
In the search for an optimal approach for the transcutaneous immunization (TCI) of hepatitis B surface antigen (HBsAg), two vesicle formulations, L595 vesicles (composed of sucrose-laurate ester and octaoxyethylene-laurate ester) and sPC vesicles (composed of soybean-phosphatidylcholine and Span-80) were prepared
Faiyaz Shakeel et al.
Drug development and industrial pharmacy, 40(9), 1240-1245 (2013-07-11)
Abstract For the development of an effective self-nanoemulsifying drug delivery system (SNEDDS) of poorly soluble drugs, the knowledge of the solubility in its oil phase and SNEDDS are one of the most important factors to avoid possibility of drug to
Daniela S Bernardi et al.
Journal of nanobiotechnology, 9, 44-44 (2011-09-29)
Nanoemulsions have practical application in a multitude of commercial areas, such as the chemical, pharmaceutical and cosmetic industries. Cosmetic industries use rice bran oil in sunscreen formulations, anti ageing products and in treatments for skin diseases. The aim of this
Keita Hayashi et al.
Colloids and surfaces. B, Biointerfaces, 106, 258-264 (2013-02-26)
We have focused on the interaction between the Span 80 vesicle and phospholipid vesicle (liposome). The Span 80 vesicle was mixed with a phospholipid vesicle (liposome), used as a simplified model of plasma membrane. From calcein leakage experiments, it was
Jingchun Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 28(17), 6769-6775 (2012-04-06)
The effects of salt on emulsions containing sorbitan oleate (Span 80) and Laponite particles were investigated. Surprisingly, a novel double phase inversion was induced by simply changing the salt concentration. At fixed concentration of Laponite particles in the aqueous phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service