Skip to Content
Merck
All Photos(1)

Key Documents

W302805

Sigma-Aldrich

Sorbitan monostearate

meets FCC analytical specifications

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46O6
CAS Number:
Molecular Weight:
430.62
FEMA Number:
3028
Beilstein:
6820761
EC Number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

reg. compliance

FDA 21 CFR 117
FDA 21 CFR 172.515
meets analytical specifications of FCC

mp

54-57 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O

InChI

1S/C24H46O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h20-21,23-26,28H,2-19H2,1H3/t20-,21+,23+,24+/m0/s1

InChI key

HVUMOYIDDBPOLL-XWVZOOPGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sorbitan monostearate is a hydrophobic nonionic surfactant is commonly used as an emulsifier, stabilizer and defoaming agent. It is also used to rehydrate yeast.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 91-91 (2012)
Pablo García-Manrique et al.
Nanomaterials (Basel, Switzerland), 10(5) (2020-05-24)
Quantitative detection of exosomes in bio-fluids is a challenging task in a dynamic research field. The absence of a well-established reference material (RM) for method development and inter-comparison studies could be potentially overcome with artificial exosomes: lab-produced biomimetic particles with
Mengmeng Shang et al.
Ultrasonics, 99, 105947-105947 (2019-07-10)
Nanobubbles (NBs) have received wide attention as theranostic agents and been extensively explored in various applications, especially in cancer. The aim of this study was to develop a novel kind of NBs which possess high echogenicity and good stability. This
Noha Ibrahim Elsherif et al.
AAPS PharmSciTech, 18(2), 551-562 (2016-05-04)
Treating a nail infection like onychomycosis is challenging as the human nail plate acts as a formidable barrier against all drug permeation. Available oral and topical treatments have several setbacks. Terbinafine hydrochloride (TBH), belonging to the allylamine class, is mainly
Ghada Ahmed El-Emam et al.
International journal of nanomedicine, 15, 7825-7840 (2020-10-30)
Voriconazole (VRC) is a triazole broad spectrum antifungal drug, used in the management of versatile fungal infections, particularly fungal keratitis. The obligatory use of niosomal delivery of VRC may reduce the frequency of dosing intervals resulting from its short biological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service