Skip to Content
Merck
All Photos(1)

Key Documents

R8005

SAFC

RPMI-1640 Medium

Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma

Pharma Manufacturing

Synonym(s):

RPMI 1640 Culture Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin

components

phenol red: 0.0053 g/L
L-glutamine: 0.3 g/L
HEPES: 3.5745 g/L
NaHCO3: no
glucose: 4.5 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Quantity

Formulated to contain 16.4 grams of powder per liter of medium.

Reconstitution

Supplement with 2.0 g/L sodium bicarbonate.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryoichi Arita et al.
Diabetes, 58(1), 215-226 (2008-10-09)
Leukocyte adhesion in retinal microvasuculature substantially contributes to diabetic retinopathy. Involvement of the Rho/Rho kinase (ROCK) pathway in diabetic microvasculopathy and therapeutic potential of fasudil, a selective ROCK inhibitor, are investigated. Localization of RhoA/ROCK and Rho activity were examined in
Tamara Veiga-Parga et al.
PLoS pathogens, 7(12), e1002427-e1002427 (2011-12-17)
Ocular herpes simplex virus infection can cause a blinding CD4⁺ T cell orchestrated immuno-inflammatory lesion in the cornea called Stromal Keratitis (SK). A key to controlling the severity of SK lesions is to suppress the activity of T cells that
Fhernanda R Smiderle et al.
BMC complementary and alternative medicine, 11, 58-58 (2011-07-27)
Mushroom polysaccharides have traditionally been used for the prevention and treatment of a multitude of disorders like infectious illnesses, cancers and various autoimmune diseases. Crude mushroom extracts have been tested without detailed chemical analyses of its polysaccharide content. For the
Yoichi Matsuo et al.
International journal of cancer, 124(4), 853-861 (2008-11-28)
CXC-chemokines are involved in the chemotaxis of neutrophils, lymphocytes and monocytes. However, role of these chemokines in tumorigenesis, especially with regard to interaction between tumor and its microenvironment, has not been clearly elucidated. The purpose of this study was to
David-Alexandre Trégouet et al.
Diabetes, 57(10), 2843-2850 (2008-07-18)
Genetic and environmental factors modulate the susceptibility to diabetic nephropathy, as initiating and/or progression factors. The objective of the European Rational Approach for the Genetics of Diabetic Complications (EURAGEDIC) study is to identify nephropathy susceptibility genes. We report molecular genetic

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Questions

  1. How to reconstitute the 10L pack for Cat#R8005-10L

    1 answer
    1. According to the COA the solubility of this item should be 16.4g/L of water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service