Skip to Content
Merck
All Photos(1)

Documents

R0883

Sigma-Aldrich

RPMI-1640 Medium

With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

sodium pyruvate: no
phenol red: yes
L-glutamine: no
NaHCO3: yes
HEPES: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Application

RPMI-1640 Medium has been used:
  • to culture PC-3 prostate cell line and Jurkat cells
  • to maintain a triple-negative MDA-MB-231 and a luminal type MCF7 human Caucasian breast adenocarcinoma cell line
  • to cultivate circulating tumor cells (CTCs)

Reconstitution

Supplement with 0.3 g/L L-glutamine.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mathur et al.
JNMA; journal of the Nepal Medical Association, 53(198), 108-112 (2016-03-20)
Dermatophytosis is a common superficial fungal infection of the skin, hair and nails caused by Trichophyton, Microsporum and Epidermophyton dermatophytic species. Identification of causative dermatophytic species tend to vary with time and place and antifungal sensitivity is of epidemiological concern
MUC1 antigen-specific CD8 T lymphocytes targeting MCF7 and MDA-MB-231 human breast adenocarcinoma cell lines
Parsonidis P, et al.
Journal of Cancer Therapy, 10(07), 495-495 (2019)
Stepan Coufal et al.
Cells, 8(7) (2019-07-25)
Crohn's disease (CD), ulcerative colitis (UC) and inflammatory bowel disease (IBD) associated with primary sclerosing cholangitis (PSC-IBD), share three major pathogenetic mechanisms of inflammatory bowel disease (IBD)-gut dysbiosis, gut barrier failure and immune system dysregulation. While clinical differences among them
Grzegorz Sumara et al.
Cell, 136(2), 235-248 (2009-01-13)
Dysfunction and loss of insulin-producing pancreatic beta cells represent hallmarks of diabetes mellitus. Here, we show that mice lacking the mitogen-activated protein kinase (MAPK) p38delta display improved glucose tolerance due to enhanced insulin secretion from pancreatic beta cells. Deletion of
Adela Ngwewondo et al.
PLoS neglected tropical diseases, 12(6), e0006565-e0006565 (2018-06-14)
Ivermectin is the only drug currently recommended for the treatment of onchocerciasis, the second leading infectious cause of blindness in the world. This drug kills only the first stage larvae-microfilariae (mf) of Onchocerca volvulus and is to be used cautiously

Protocols

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

WST-1 assay protocol for cell viability and cytotoxicity measurements with preparation instructions, applications, FAQs, and troubleshooting.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service