Skip to Content
Merck
All Photos(1)

Key Documents

R8755

Sigma-Aldrich

RPMI-1640 Medium

With ʟ-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Roswell Park Memorial Institute 1640 medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture

form

powder

technique(s)

cell culture | mammalian: suitable

components

HEPES: no
phenol red: no
L-glutamine: yes
NaHCO3: no
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage-dependent. Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum. It has also been widely used in fusion protocols and in the growth of hybrid cells. This medium is suitable for culturing human normal and neoplastic leukocytes.

Application

RPMI-1640 Medium is used for the following applications:
  • Used in cell culture medium: Murine thymoma EL-4 cells were cultured in RPMI 1640 medium supplemented with other components
  • Used for the mycobacterial infection of splenocytes (in antibiotic-free RPMI 1640 medium)
  • Used for cell isolation and culture
  • Used in culture medium (as one of the component) during lymphocyte separation and culture, ELISPOT Assay, Ex Vivo Proliferation Assay
  • Used in medium for gp39 gene expression and CD40-Immunoglobuin binding assays
  • Used for the preparation of antifungal agents

Quantity

Formulated to contain 10.4 grams of powder per liter of medium.

Reconstitution

Supplement with 2.0 g/L sodium bicarbonate.

Other Notes

Phenol red has been shown to interfere with the growth of some cells at clonal densities. Use this medium when working with stem cells or when growing cells at low densities. Also recommended for in vitro diagnostics use.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R Roubenoff et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 53(1), M20-M26 (1998-02-19)
To determine the association among aging, inflammation, and cytokine production by peripheral blood mononuclear cells. We examined production of interleukin-1 beta (IL-1 beta), tumor necrosis factor-alpha (TNF-alpha), IL-1 receptor antagonist (IL-1Ra), and IL-6 in 711 elderly participants in the Framingham
Manuel Cuenca-Estrella et al.
Antimicrobial agents and chemotherapy, 46(11), 3644-3647 (2002-10-18)
A two-laboratory study was performed to evaluate the correlation between the NCCLS M27-A and EUCAST microdilution procedures for antifungal testing of Candida spp. A panel of 109 bloodstream isolates was tested against amphotericin B, flucytosine, fluconazole, and itraconazole. Overall, the
Mathias Lichterfeld et al.
The Journal of experimental medicine, 200(6), 701-712 (2004-09-24)
Virus-specific CD8(+) T cells are associated with declining viremia in acute human immunodeficiency virus (HIV)1 infection, but do not correlate with control of viremia in chronic infection, suggesting a progressive functional defect not measured by interferon gamma assays presently used.
T P Szatrowski et al.
Cancer research, 51(3), 794-798 (1991-02-01)
Few nonphagocytic cells are known to generate reactive oxygen intermediates. Based on horseradish peroxidase-dependent, catalase-inhibitable oxidation of fluorescent scopoletin, seven human tumor cell lines constitutively elaborated H2O2 at rates (up to 0.5 nmol/10(4) cells/h) large enough that cumulative amounts at
M E Klepser et al.
Antimicrobial agents and chemotherapy, 41(6), 1392-1395 (1997-06-01)
Time-kill curves were determined for three isolates of Candida albicans tested against fluconazole and amphotericin B at multiples of the MIC. Fluconazole produced fungistatic activity, with concentration-related growth effects observed over a narrow range of concentrations. Amphotericin B exhibited fungicidal

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Protocols

Powdered media and salt mixtures are extremely hygroscopic and should be protected from atmospheric moisture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service