Skip to Content
Merck
All Photos(1)

Documents

K4144

Sigma-Aldrich

KB-R7943

≥98% (HPLC), powder

Synonym(s):

2-[4-[(4-nitrophenyl)methoxy]phenyl]ethyl ester, methanesulfonate (1:1), Carbamimidothioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N3O3S·CH3SO3H
CAS Number:
Molecular Weight:
427.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to tan

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.NC(=N)SCCc1ccc(OCc2ccc(cc2)[N+]([O-])=O)cc1

InChI

1S/C16H17N3O3S.CH4O3S/c17-16(18)23-10-9-12-3-7-15(8-4-12)22-11-13-1-5-14(6-2-13)19(20)21;1-5(2,3)4/h1-8H,9-11H2,(H3,17,18);1H3,(H,2,3,4)

InChI key

WGIKEBHIKKWJLG-UHFFFAOYSA-N

Biochem/physiol Actions

KB-R7943 inhibits the reversed Na(+)/Ca(2+) exchanger, NCX. In cardiomyocytes, Ca2+ is released from intracellular stores during contraction, and sequestered again during relaxation. NCX is the primary mechanism that prevents Ca2+ overload due to influx of calcium across the plasma membrane.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignacio Ibáñez et al.
Neurochemistry international, 123, 125-132 (2018-03-27)
GLT-1 is the main glutamate transporter in the brain and its trafficking controls its availability at the cell surface, thereby shaping glutamatergic neurotransmission under physiological and pathological conditions. Extracellular glutamate is known to trigger ubiquitin-dependent GLT-1 internalization from the surface
Camille Loubiere et al.
Scientific reports, 7(1), 5040-5040 (2017-07-13)
Mitochondrial integrity is critical for the regulation of cellular energy and apoptosis. Metformin is an energy disruptor targeting complex I of the respiratory chain. We demonstrate that metformin induces endoplasmic reticulum (ER) stress, calcium release from the ER and subsequent
Reno C Reyes et al.
ASN neuro, 4(1) (2012-01-25)
Astroglial excitability operates through increases in Ca2+cyt (cytosolic Ca2+), which can lead to glutamatergic gliotransmission. In parallel fluctuations in astrocytic Na+cyt (cytosolic Na+) control metabolic neuronal-glial signalling, most notably through stimulation of lactate production, which on release from astrocytes can
Sandra Mitrovic et al.
eLife, 2, e00658-e00658 (2013-06-07)
Mucin 5AC (MUC5AC) is secreted by goblet cells of the respiratory tract and, surprisingly, also expressed de novo in mucus secreting cancer lines. siRNA-mediated knockdown of 7343 human gene products in a human colonic cancer goblet cell line (HT29-18N2) revealed
Lorena Halty-deLeon et al.
Frontiers in cellular neuroscience, 12, 186-186 (2018-07-19)
CALX, the Na+/Ca2+ exchanger in Drosophila, is highly expressed in the outer dendrites of olfactory sensory neurons (OSNs) which are equipped with the odorant receptors (ORs). Insect OR/Orco dimers are nonselective cation channels that pass also calcium which leads to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service