Skip to Content
Merck
All Photos(5)

Key Documents

N3501

Sigma-Aldrich

β-Nicotinamide mononucleotide

≥95% (HPLC)

Synonym(s):

β-NMN, β-Nicotinamide ribose monophosphate, NMN, Nicotinamide ribotide, Nicotinamide-1-ium-1-β-D-ribofuranoside 5′-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N2O8P
CAS Number:
Molecular Weight:
334.22
Beilstein:
3570187
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥95% (HPLC)
≥95% (spectrophotometric assay)

form

powder

solubility

H2O: soluble-50 mg/mL to clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

C[C@H]1[C@@H](C)[C@@H](O[C@@H]1COP(O)([O-])=O)[n+]2cccc(c2)C(N)=O

InChI

1S/C13H19N2O6P/c1-8-9(2)13(21-11(8)7-20-22(17,18)19)15-5-3-4-10(6-15)12(14)16/h3-6,8-9,11,13H,7H2,1-2H3,(H3-,14,16,17,18,19)/t8-,9+,11+,13+/m0/s1

InChI key

YYNOOWWEYJELSF-SEMCEJNYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Nicotinamide mononucleotide (β-NMN) is an intermediate in the nicotinamide phosphoribosyltransferase (NAMPT)-catalyzed biosynthesis of nicotinamide adenine dinucleotide (NAD+). NAMPT mediates the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to produce β-NMN. β-NMN adenyltransferase subsequently converts β-NMN to NAD+.

Application

β-Nicotinamide mononucleotide (NMN) is used to study binding motifs within RNA aptamers and ribozyme activation processes involving β-nicotinamide mononucleotide (β-NMN)-activated RNA fragments.

Other Notes

Note: This is the common form of NMN. Do not confuse it with α-NMN.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C T Lauhon et al.
Journal of the American Chemical Society, 117(4), 1246-1257 (1995-02-01)
RNA molecules that specifically bind riboflavin (Rb) and beta-nicotinamide mononucleotide (NMN) have been isolated by in vitro selection. A simple structural motif containing intramolecular G-quartets was found to bind tightly to oxidized riboflavin (Kd = 1-5 micromolar). DNA versions of
João Meireles Ribeiro et al.
Scientific reports, 8(1), 1036-1036 (2018-01-20)
Cyclic ADP-ribose (cADPR) is a messenger for Ca
Zheng-Hu Shi et al.
Scientific reports, 7(1), 8041-8041 (2017-08-16)
Metanephrines (MNs) were suggested as a potential first-line biochemical index for the diagnosis of phaeochromocytomas (PHEO). In this study, we developed a simple electrochemical method for the quantitative measurement of MNs in spot urine samples. As MNs contain a hydroxyphenyl
Sophia E Airhart et al.
PloS one, 12(12), e0186459-e0186459 (2017-12-07)
The co-primary objectives of this study were to determine the human pharmacokinetics (PK) of oral NR and the effect of NR on whole blood nicotinamide adenine dinucleotide (NAD+) levels. Though mitochondrial dysfunction plays a critical role in the development and
Kazuhiro Hasegawa et al.
Nature medicine, 19(11), 1496-1504 (2013-10-22)
Sirtuin 1 (Sirt1), a NAD(+)-regulated deacetylase with numerous known positive effects on cellular and whole-body metabolism, is expressed in the renal cortex and medulla. It is known to have protective effects against age-related disease, including diabetes. Here we investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service