Skip to Content
Merck
All Photos(1)

Documents

F8636

Sigma-Aldrich

Ficoll® 400

lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture

Synonym(s):

Polysucrose 400

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.75

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

Looking for similar products? Visit Product Comparison Guide

Application

Ficoll is a hydrophilic neutral highly branched polysaccharide use to establish density gradients to separate blood component cells and organelles. It is part of the separation medium called Ficoll-Paque.
Used for cell separation and organelle isolation.

Other Notes

A nonionic synthetic polymer of sucrose.
Dialyzed

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yann Peytour et al.
Transfusion, 50(10), 2152-2157 (2010-05-12)
Human CD34+ cells are mandatory to study many aspects of human hematopoiesis. Their low frequency in blood or marrow and ethical reasons limit their obtainment in large quantities. Leukoreduction filters (LRFs) are discarded after preparation of red blood cells. The
A Schoug et al.
Journal of applied microbiology, 108(3), 1032-1040 (2009-09-09)
To investigate the effect of freeze-dried Lactobacillus coryniformis Si3 on storage stability by adding polymers to sucrose-based formulations and to examine the relationship between amorphous matrix stability and cell viability. The resistance to moisture-induced sucrose crystallization and effects on the
Weihua Fei et al.
PLoS genetics, 7(7), e1002201-e1002201 (2011-08-11)
Lipid droplets (LDs) are important cellular organelles that govern the storage and turnover of lipids. Little is known about how the size of LDs is controlled, although LDs of diverse sizes have been observed in different tissues and under different
Louise A Knight et al.
BMC cancer, 9, 38-38 (2009-01-30)
Previous data suggest that lipophilic statins such as fluvastatin and N-bisphosphonates such as zoledronic acid, both inhibitors of the mevalonate metabolic pathway, have anti-cancer effects in vitro and in patients. We have examined the effect of fluvastatin alone and in
Naoaki Sakata et al.
Transplantation, 87(6), 825-830 (2009-03-21)
There is a recent focus on embolization of the portal vein by transplanted islets as a major cause of early graft loss. The resultant ischemia causes necrosis or apoptosis of cells within the liver. Thus, noninvasive assessment of the liver

Protocols

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service