Skip to Content
Merck
All Photos(1)

Key Documents

250233

Sigma-Aldrich

Cytochalasin B

from Helminthosporium dematioideum, ≥98% (HPLC), solid, cell-permeable fungal toxin, Calbiochem®

Synonym(s):

Cytochalasin B, Helminthosporium dematioideum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37NO5
CAS Number:
Molecular Weight:
479.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Cytochalasin B, Helminthosporium dematioideum, Cell-permeable fungal toxin.

Quality Level

description

Merck USA index - 14, 2790

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

methanol: 20 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N1[C@H]([C@H]3[C@@]4(OC(=O)\C=C\[C@@H](CCC[C@H](C\C=C\[C@H]4[C@@H](C(=C)[C@H]3C)O)C)O)C1=O)Cc2ccccc2

InChI

1S/C29H37NO5/c1-18-9-7-13-22(31)15-16-25(32)35-29-23(14-8-10-18)27(33)20(3)19(2)26(29)24(30-28(29)34)17-21-11-5-4-6-12-21/h4-6,8,11-12,14-16,18-19,22-24,26-27,31,33H,3,7,9-10,13,17H2,1-2H3,(H,30,34)/b14-8+,16-15+/t18-,19-,22-,23+,24+,26+,27-,29-/m1/s1

InChI key

GBOGMAARMMDZGR-TYHYBEHESA-N

General description

Cell-permeable fungal toxin. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments. Inhibits cell movement and induces nuclear extrusion. Shortens actin filaments by blocking monomer addition at the fast growing end of polymers. Inhibits glucose transport and platelet aggregation. Blocks adenosine-induced apoptotic body formation without affecting activation of endogenous ADP-ribosylation in leukemia HL-60 cells.
Cell-permeable fungal toxin. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments. Inhibits cell movement and induces nuclear extrusion. Shortens actin filaments by blocking monomer addition at the fast-growing end of polymers. Inhibits glucose transport and platelet aggregation. Blocks adenosine-induced apoptotic body formation without affecting activation of endogenous ADP-ribosylation in leukemia HL-60 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Blocking the formation of contractile microfilaments
Product does not compete with ATP.
Reversible: no

Caution

Poor quality DMSO will cause degradation of the product, so high quality DMSO is recommended.

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following reconstitution store in the refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Tanaka, Y., et al. 1994. Exp. Cell Res. 213, 242.
Theodoropoulos, P.A., et al. 1994. Biochem. Pharmacol. 47, 1875.
Benya, P.D., and Padilla, S.R. 1993. Exp. Cell Res. 204, 268.
Cairns, M.T., et al. 1991. J. Biol. Chem.266, 8176.
Nowak, D. 1990. Biomed. Biochim. Acta49, 353.
Wang, F., et al. 1990. Biochem. Biophys. Res. Commun.171, 543.
Edwards, S.W., and Lloyd, D. 1988. FEBS Lett.227, 39.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Meng et al.
Frontiers in cell and developmental biology, 9, 746237-746237 (2021-11-13)
[Ca2+]i is essential for mammalian oocyte maturation and early embryonic development, as those processes are Ca2+ dependent. In the present study, we investigated the effect of [Ca2+]i on in vitro maturation and reprogramming of oocytes in a lower calcium model
Shuyue Qi et al.
PLoS biology, 20(8), e3001741-e3001741 (2022-08-17)
Mitochondrial DNA (mtDNA) mutations are often associated with incurable diseases and lead to detectable pathogenic variants in 1 out of 200 babies. Uncoupling of the inheritance of mtDNA and the nuclear genome by spindle transfer (ST) can potentially prevent the
Naomi Watanabe et al.
Scientific reports, 13(1), 11175-11175 (2023-07-11)
Wild-derived mouse strains have been extensively used in biomedical research because of the high level of inter-strain polymorphisms and phenotypic variations. However, they often show poor reproductive performance and are difficult to maintain by conventional in vitro fertilization and embryo
Michael J Burgum et al.
Mutagenesis, 39(3), 205-217 (2024-03-19)
The current Organisation for Economic Co-Operation and Development test guideline number 487 (OECD TG No. 487) provides instruction on how to conduct the in vitro micronucleus assay. This assay is one of the gold standard approaches for measuring the mutagenicity
Jing Chen et al.
Scientific reports, 11(1), 16798-16798 (2021-08-20)
Tunneling nanotube (TNT), a dynamic cell-cell contact, is dependent on actin polymerization. TNTs are efficient in transporting ions, proteins and organelles intercellularly, which are important mechanisms in physiological and pathological processes. Reported studies on the existence and function of TNTs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service