Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ficoll
®
400
Synonym(s):
Polysucrose 400
CAS Number:
26873-85-8
Products
Product Category
carbohydrates (1)
polysaccharides (1)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
F2637
Ficoll
®
400,
BioXtra, Molecular Biology, lyophilized powder
Expand
View Pricing
F9378
Ficoll
®
400,
Expand
View Pricing
F8016
Ficoll
®
400,
lyophilized powder, Type 400-DL, BioReagent, suitable for cell culture
Expand
View Pricing
F1418
Ficoll
®
400,
BioXtra, Type 400-DL, lyophilized powder
Expand
View Pricing
F8636
Ficoll
®
400,
lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Expand
View Pricing