Skip to Content
Merck
All Photos(2)

Documents

F1418

Sigma-Aldrich

Ficoll® 400

BioXtra, Type 400-DL, lyophilized powder

Synonym(s):

Polysucrose 400

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

type

Type 400-DL

product line

BioXtra

form

lyophilized powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

<0.1%

color

white

solubility

water: soluble

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Application

Ficoll® 400 is used together with sodium diatrizoate (Paque) to create Ficoll-Paque, a density mixture, used for the separation of cells and organelles, especially lymphocytes and monocytes from blood. Ficoll 400® may be used for the development of new cell and organelle separation systems. Ficoll 400 is used in tissue engineering and macromolecular conformation research for the development, evaluation and use of macromolecular crowding (MMC) systems and configurations.
Used for cell separation and organelle isolation.

Other Notes

A nonionic synthetic polymer of sucrose.
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soluble form of the endothelial adhesion molecule CD146 binds preferentially CD16+ monocytes.
Garibaldi S, Barisione C, Ghigliotti G, et al.
Molecular Biology Reports, 39(6), 6745-6752 (2011)
T Min et al.
Transplantation proceedings, 42(5), 1825-1829 (2010-07-14)
Sufficient and favorable biological functions of islets are major problems hindering xenotransplantation. The aim of the present study was to evaluate the effects on harvesting, purity, viability, and function of using improved Visipaque (iodixanol) and Ficoll-400 for adult porcine islet
Selective Expansion of CD34+ Cells from Mouse Bone Marrow Cultured on LH/P MP-Coated Plates with Adequate Cytokines.
Kishimoto S, Ishihara M, Kanatani Y, et al.
Journal of Tissue Engineering, 2, doi: 10-doi: 10 (2011)
Yanyan Xu et al.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 24(4), 406-409 (2010-05-13)
The purity and activity of islets will greatly affect the outcome of xenotransplantation therapy of type 1 diabetes mellitus. To set up an improved method of the isolation and purification of rat islets, which can obtain high-purity, high-yield, and high-viability
Bo Chen et al.
Tissue engineering. Part C, Methods, 19(8), 586-595 (2013-02-21)
Macromolecular crowding (MMC) has been shown to have a beneficial effect on production and maturation of extracellular matrices in a monolayer cell culture model. To explore its potential in tissue engineering, a mixture of Ficoll 70 and Ficoll 400 is

Protocols

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Ficoll 400® is a non-ionic polymer with high hydrophilicity and water solubility, suitable for various biological applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service