Skip to Content
Merck
All Photos(2)

Key Documents

E0127

Sigma-Aldrich

Elastase from porcine pancreas

Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein

Synonym(s):

Elastase from hog pancreas, Pancreatopeptidase E

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type III

form

lyophilized powder

specific activity

≥4.0 units/mg protein

mol wt

25.9 kDa

composition

Protein, 55-85%

solubility

200 mM Tris HCl buffer, pH 8.8: soluble 1.0 mg/mL, clear

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Elastase from porcine pancreas has been used in a study to identify a novel bone/calcium metabolism-regulating factor in porcine pancreas. Elastase from porcine pancreas has also been used in a study to investigate the effect of a specific synthetic inhibitor of neutrophil elastase (ONO-5046) on the course of acute hemorrhagic pancreatitis in dogs.
Elastase from porcine pancreas has been used in the proteolytic cleavage of UMP synthase (Uridine-5′-monophosphate synthase). It has also been used along with collagenase to disperse tissue into single cells from the thoracic media, abdominal aortic media and intima of rabbit.

Biochem/physiol Actions

Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.
Elastase is a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges. The molecular mass is approximately 25.9 kDa. The enzyme is synthesized as an inactive zymogen, proelastase, which is converted to the active form by limited proteolysis at the N-terminal by trypsin. It is a serine protease with broad specificity. It cleaves protein at the carboxyl side of small hydrophobic amino acids such as Ile, Gly, Ala, Ser, Val, and Leu. The enzyme also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester. The pH optimum is found to be 8.0-8.5. It does not require any activator, but it is inhibited by diisopropyl fluorophosphate, phenylmethanesulfonyl fluoride, α2-macroglobulin, α1-antitrypsin, sulfonyl fluorides, p-dinitrophenyl diethylphosphate and high salt concentrations. Elastase is extensively used in tissue and cell dissociation procedures. It is effective in the isolation of Type II lung cells.

Packaging

Package size based on protein content

Quality

Contains trypsin activity

Unit Definition

One unit will hydrolyze 1.0 μmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per min, pH 8.0 at 25 °C.

Physical form

Contains sodium carbonate.

Preparation Note

2x crystallized and chromatographically purified

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R H Kelly et al.
Digestion, 22(1), 1-7 (1981-01-01)
Induction of acute hemorrhagic pancreatic necrosis by dietary means in mice produces alterations in the serum complement system. Total hemolytic complement, i.e., CH50, and native C3 levels fall during the development of pancreatitis while, at the same time, what could
J H Campbell et al.
Arteriosclerosis (Dallas, Tex.), 5(4), 318-328 (1985-07-01)
Smooth muscle cells of the rabbit aorta, when grown in vitro, express three distinguishable forms of phenotype (contractile, reversible synthetic, and irreversible synthetic). We compared the interactions of these three smooth muscle phenotypes with rabbit very low density lipoprotein (VLDL)
E E Floyd et al.
The Journal of biological chemistry, 260(16), 9443-9451 (1985-08-05)
The multifunctional protein uridine 5'-monophosphate (UMP) synthase catalyzes the final two reactions of the de novo biosynthesis of UMP in mammalian cells by the sequential action of orotate phosphoribosyltransferase (EC 2.4.2.10) and orotidine 5'-monophosphate (OMP) decarboxylase (EC 4.1.1.23). This protein
Ting Wang et al.
Biochemical and biophysical research communications, 485(2), 284-289 (2017-02-23)
Acute lung injury (ALI) is an early pathophysiologic change in acute respiratory distress syndrome and its management can be challenging. Omalizumab (Xolair™) is a recombinant DNA-derived, humanized antibody. OMZ-SPT is a polypeptide on the heavy chain of omalizumab monoclonal antibody.
Domenico M Guarascio et al.
The Journal of physiology, 599(15), 3697-3714 (2021-06-06)
Taste transduction occurs in taste buds in the tongue epithelium. The Ca2+ -activated Cl- channels TMEM16A and TMEM16B play relevant physiological roles in several sensory systems. Here, we report that TMEM16A, but not TMEM16B, is expressed in the apical part

Protocols

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service