Skip to Content
Merck
All Photos(2)

Documents

E7885

Sigma-Aldrich

Elastase from porcine pancreas

lyophilized powder, suitable for cell culture

Synonym(s):

Elastase from hog pancreas, Pancreatopeptidase E

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

biological source

Porcine

form

lyophilized powder

specific activity

≥4 units/mg protein

mol wt

25.9 kDa

packaging

pkg of 1 μmol

technique(s)

cell culture | mammalian: suitable

pH

8

suitability

suitable for

foreign activity

Trypsin ≤50 BAEE units/mg protein

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Elastase is a single polypeptide chain of 240 amino acid residues and contains four disulfide bridges. The molecular mass is approximately 25.9kDa. The enzyme is synthesized as an inactive zymogen, proelastase, which is converted to the active form by limited proteolysis at the N-terminal by trypsin.

Application

Elastase from porcine pancreas is used for the following applications:
  • Cell culture techniques
  • ELISA techniques
  • Used in assay for elastase inhibitory activity

Biochem/physiol Actions

Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.
Enzyme that degrades elastin, a matrix component of tissues involved in stretching processes. It is a serine protease with broad specificity. It cleaves protein at the carboxyl side of small hydrophobic amino acids such as Ile, Gly, Ala, Ser, Val, and Leu. The enzyme also hydrolyzes amides and esters such as N-Benzoyl-L-alanine methyl ester.

Packaging

Package size based on protein content

Unit Definition

One unit will hydrolyze 1.0 μmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per min, pH 8.0 at 25 °C.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrien Lupieri et al.
Cardiovascular research, 116(2), 438-449 (2019-05-21)
Defects in efficient endothelial healing have been associated with complication of atherosclerosis such as post-angioplasty neoatherosclerosis and plaque erosion leading to thrombus formation. However, current preventive strategies do not consider re-endothelialization in their design. Here, we investigate mechanisms linking immune
P Hamet et al.
The Journal of biological chemistry, 264(21), 12364-12369 (1989-07-25)
The induction of cyclic GMP formation in target tissues, i.e. vascular smooth muscle and endothelial cells, by atrial natriuretic factor is followed by its egression into plasma and urine. Since the extracellular appearance of this cyclic nucleotide is used as
Takeshi Yanagisawa et al.
Stroke, 51(10), 3083-3094 (2020-09-12)
Intracranial aneurysm formation and rupture risk are, in part, determined by genetic factors and sex. To examine their role, we compared 3 mouse strains commonly used in cerebrovascular studies in a model of intracranial aneurysm formation and rupture. Intracranial aneurysms
Yoshiyuki Okumura et al.
Nihon Ishinkin Gakkai zasshi = Japanese journal of medical mycology, 48(1), 13-18 (2007-02-09)
The amino acid sequence of elastase inhibitor, AFLEI, isolated from Aspergillus flavus was determined by the Edman sequencing procedure of peptides derived from digests utilizing clostripain. A molecular weight of 7,525.8 was observed by TOF-MS. AFLEI contained 68 amino acid
F Seibold et al.
Gut, 32(10), 1192-1197 (1991-10-01)
The presence of antibodies against pancreatic juice (PAB) in patients with Crohn's disease has recently been reported. In our study sera from 273 patients with inflammatory bowel disease (222 with Crohn's disease, 51 with ulcerative colitis) have been examined for

Articles

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Discover pre-mixed collagenase enzyme blends with DNase I, Dispase II, Elastase, and Hyaluronidase and gently dissociate animal tissues in vitro.

Protocols

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Enzymatic Assay of Elastase (EC 3.4.21.36), a continuous spectrophotometric rate determination.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service