Skip to Content
Merck
All Photos(2)

Key Documents

C9134

Sigma-Aldrich

α-Chymotrypsin−Agarose from bovine pancreas

lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine pancreas

Quality Level

form

lyophilized powder

specific activity

2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)

mol wt

25 kDa

UniProt accession no.

storage temp.

−20°C

Gene Information

Related Categories

Application

α-Chymotrypsin agarose from bovine pancreas has been used to study the purification and characterization of glucoamylase. α-Chymotrypsin agarose from bovine pancreas has also been used in a study to investigate molecular modeling for the design of a biomimetic chimeric ligand.

Biochem/physiol Actions

α-Chymotrypsin is a serine peptidase and has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. Molecular weight of this enzyme is found to be 25 kDa. The pI is 8.75. It selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan, and leucine. Ca2+ activates and stabilizes the enzyme. The enzyme is inhibited by diisopropyl fluorophosphate (DFP), phenylmethanesulfonyl fluoride (PMSF), N-p-tosyl-L-phenylalanine chloromethyl ketone (TPCK), chymostatin, aprotinin, α1-antitrypsin, and α2-macroglobulin, 10 mM Cu2+ and Hg2+.
Serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met) on the carboxyl end of the bond.

Unit Definition

One unit will hydrolyze 1.0 μmole of N-acetyl-L-tyrosine ethyl ester (ATEE) per min at pH 8.0 at 30 °C.

Physical form

Stabilized with lactose

Analysis Note

Protein determined by A1%/280

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukiko Taketani et al.
Bioscience, biotechnology, and biochemistry, 75(10), 1872-1879 (2011-10-08)
Thioredoxin (TRX) catalyzes the reduction of disulfide bonds in proteins via the NADPH-dependent thioredoxin reductase system. Reducing the disulfide bonds of allergenic proteins in food by TRX lowers the allergenicity. We established in this study a method to prepare TRX-enriched
Saleh A Mohamed et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 140(1), 73-80 (2004-12-29)
The present study analyzed the existence of carbohydrases in camel pancreas compared to some other ruminants. Disaccharidases (maltase, cellobiase, lactase, trehalase and sucrase), glucoamylase and alpha-amylase were detected in pancreas of camel, sheep, cow and buffalo. Enzyme levels in sheep
N E Labrou et al.
Biotechnology and bioengineering, 63(3), 322-332 (1999-04-01)
Molecular modeling was employed for the design of a biomimetic chimeric ligand for L-lactate dehydrogenase (LDH). This ligand is an anthraquinone monochlorotriazinyl dye comprising two moieties: (a) the ketocarboxyl biomimetic moiety, 2-(4-aminophenyl)-ethyloxamic acid, linked on the monochlorotriazine ring, mimicking the
Ke Xu et al.
The Journal of biological chemistry, 283(49), 34337-34344 (2008-10-11)
The clinical severity of Osteogenesis Imperfecta (OI), also known as the brittle bone disease, relates to the extent of conformational changes in the collagen triple helix induced by Gly substitution mutations. The lingering question is why Gly substitutions at different
Carolina Garri et al.
Oncotarget, 9(44), 27363-27379 (2018-06-26)
The cancer-associated protein Anterior Gradient 2 (AGR2) has been described, predominantly in adenocarcinomas. Increased levels of extracellular AGR2 (eAGR2) have been correlated with poor prognosis in cancer patients, making it a potential biomarker. Additionally, neutralizing AGR2 antibodies showed preclinical effectiveness

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service