Skip to Content
Merck

A3401

Ala-Ala-Phe-7-amido-4-methylcoumarin

protease substrate

Synonym(s):

H-Ala-Ala-Phe-AMC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H28N4O5
CAS Number:
Molecular Weight:
464.51
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (TLC)

form

powder

solubility

ethanol: 20 mg/mL, clear, colorless to light yellow

storage temp.

2-8°C

SMILES string

CC(N)C(=O)NC(C)C(=O)NC(Cc1ccccc1)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C25H28N4O5/c1-14-11-22(30)34-21-13-18(9-10-19(14)21)28-25(33)20(12-17-7-5-4-6-8-17)29-24(32)16(3)27-23(31)15(2)26/h4-11,13,15-16,20H,12,26H2,1-3H3,(H,27,31)(H,28,33)(H,29,32)

InChI key

FVRLYIFIDKXFHU-UHFFFAOYSA-N

General description

Substrate for chymotrypsin and tripeptidyl peptidaseII.
Ala-Ala-Phe-7-amido-4-methylcoumarin is a fluorogenic substrate for the proteolytic activity. It is a positively charged substrate molecule.

Application

Ala-Ala-Phe-7-amido-4-methylcoumarin has been used:
  • in the preparation of the reaction mixture for enzymatic assays
  • to initiate the enzyme reaction in tripeptidyl peptidase-1 (TPP1) enzyme activity assay
  • to initiate the assay reaction in lysosomal hydrolases activity assay



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library