Skip to Content
Merck
All Photos(4)

Key Documents

C4879

Sigma-Aldrich

α-Chymotrypsinogen A from bovine pancreas

essentially salt-free, lyophilized powder

Synonym(s):

chymotrypsin A zymogen

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine pancreas

type

Type II

form

essentially salt-free, lyophilized powder

specific activity

≥40 units/mg solid

mol wt

25,656 Da by calculation

purified by

6× crystallization

solubility

1 mM HCl: soluble 10 mg/mL, clear, colorless

UniProt accession no.

foreign activity

α-chymotrypsin ≤1 U/mg (prior to activation by trypsin)

storage temp.

−20°C

Gene Information

Related Categories

General description

Chymotrypsinogen from bovine pancreas is a zymogen containing 5 disulfide bridges. It has an isoelectric pH of 8.97.

Application

α-Chymotrypsinogen A from bovine pancreas has been used as model protein crystallization reproducibility studies. It has also been used in the hydrolysis of α-gliadins prior to mass spectroscopy studies.
The enzyme from Sigma has been used in the non-invasive determination of solid-state protein conformation using near infrared (NIR) spectroscopy. It has been used to study the partitioning of protein in polymer/polymer aqueous two-phase systems. The enzyme has also been used for self-interaction chromatography applications, to test the rapid measurement of protein osmotic second virial coefficients. In this technique, the protein is immobilized on chromatographic particles and its retention is measured using isocratic elution.

Biochem/physiol Actions

Chymotrypsinogen A requires limited proteolysis for its activation. Chymotrypsinogen A may be activated by trypsin and chymotrypsin (autolytic activation) to form m α, β, γ, δ and π chymotrypsin (depending upon the conditions of activation). Chymotrypsin is a protease that will preferentially cleave peptides on the carboxyl side of aromatic amino acids including tryptophan, tyrosine, and phenylalanine. It will also hydrolyze peptides on the carboxyl side of leucine, methionine, and alanine.
A serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met) on the carboxyl end of the peptide bond.

Unit Definition

After activation to Chymotrypsin, one unit will hydrolyze 1.0 μmole of BTEE per min at pH 7.8 at 25 °C.

Other Notes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the activation of bovine chymotrypsinogen A. Conformational isomerization of alpha1-and kappa-chymotrypsin and their autolytic conversion to alpha-and gamma-chymotrypsin.
Sharma SK and Hopkins TR
The Journal of Biological Chemistry, 253(9), 3055-3061 (1978)
Structural basis of IgE binding to alpha-and gamma-gliadins: Contribution of disulfide bonds and repetitive and nonrepetitive domains
Mameri H, et al.
Journal of Agricultural and Food Chemistry, 63(29), 6546-6554 (2015)
9.1 Proteases: Facilitating a Difficult Reaction
Biochemistry (5th Edition) (2002)
Peter M Tessier et al.
Biophysical journal, 82(3), 1620-1631 (2002-02-28)
Weak protein interactions are often characterized in terms of the osmotic second virial coefficient (B(22)), which has been shown to correlate with protein phase behavior, such as crystallization. Traditional methods for measuring B(22), such as static light scattering, are too
Simulation of the activation of alpha-chymotrypsin: Analysis of the pathway and role of the propeptide
Matrai J, et al.
Protein Science, 13(12), 3139-3150 (2004)

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service