Skip to Content
Merck
All Photos(1)

Key Documents

C7938

Sigma-Aldrich

2-Chloro-N6-cyclopentyladenosine

adenosine receptor agonist

Synonym(s):

CCPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20ClN5O4
CAS Number:
Molecular Weight:
369.80
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

methanol: 19.60-20.40 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NC4CCCC4)nc(Cl)nc23

InChI

1S/C15H20ClN5O4/c16-15-19-12(18-7-3-1-2-4-7)9-13(20-15)21(6-17-9)14-11(24)10(23)8(5-22)25-14/h6-8,10-11,14,22-24H,1-5H2,(H,18,19,20)/t8-,10-,11-,14-/m1/s1

InChI key

XSMYYYQVWPZWIZ-IDTAVKCVSA-N

Application

2-Chloro-N6-cyclopentyladenosine (CCPA) may be used as an A1 adenosine receptor agonist with high selectivity. CCPA is often used with other adenosine receptor agonists and antagonist to resolve A(1), A(2A) and A(3) receptor functions. These include DPCPX (A(1) receptor antagonist), 4-(2-[7-amino-2-(2-furyl)[1,2,4]triazolo[2,3-α][1,3,5]triazin-5-ylamino]ethyl)phenol (A(2A) receptor antagonist), and 4-[2-[[6-amino-9-(N-ethyl-β-D-ribofuranuronamidosyl)-9H-purin-2-yl]amino]ethyl]benzenepropanoic acid hydrochloride (A(2A) receptor agonist), and Cl-IB-MECA, (A(3)) receptor agonist).

Biochem/physiol Actions

A1 adenosine receptor agonist with high selectivity: reportedly 10,000-fold for A1 over A2 receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N L D Smith et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 20(8), O480-O488 (2013-11-28)
Chronic cavitary pulmonary aspergillosis (CCPA) is a progressive lung condition with a 10-30% annual mortality. Although overtly immunocompetent, some immunogenetic defect in patients is likely. To investigate a possible immunogenetic defect in CCPA, we analysed biologically plausible candidate genes in
Romeo Romagnoli et al.
Journal of medicinal chemistry, 51(18), 5875-5879 (2008-08-30)
The synthesis and evaluation of a series of 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene derivatives as allosteric enhancers of the A 1-adenosine receptor are described. The nature of substituents on the phenyl ring tethered to the piperazine seem to exert a fundamental influence on the
Mary Clare Luca et al.
Clinical hemorheology and microcirculation, 49(1-4), 287-293 (2012-01-05)
Despite decades of research and thousands of experimental publications, acute preconditioning strategies have yet to be implemented in clinical practice. While some have attributed this to a failure of the experimental studies to mimic the clinical environment, others have suggested
Ruzanna Shkhyan et al.
Journal of molecular medicine (Berlin, Germany), 96(10), 1049-1060 (2018-08-09)
Osteoarthritis (OA), the most common form of arthritis, is characterized by inflammation of joints and cartilage degradation leading to disability, discomfort, severe pain, inflammation, and stiffness of the joint. It has been shown that adenosine, a purine nucleoside composed of
Ellen V Langemeijer et al.
Purinergic signalling, 9(1), 91-100 (2012-10-12)
The concept of functional selectivity offers great potential for the development of drugs that selectively activate a specific intracellular signaling pathway. During the last few years, it has become possible to systematically analyse compound libraries on G protein-coupled receptors (GPCRs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service