Skip to Content
Merck
All Photos(4)

Key Documents

A2647

Sigma-Aldrich

Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate

≥93% (HPLC), powder

Synonym(s):

β,γ-Imidoadenosine 5′-triphosphate lithium salt hydrate, AMP-PNP, ATP[β,γ-NH], Adenylyl imidodiphosphate lithium salt hydrate, App(NH)p

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N6O12P3 · xLi+ · yH2O
CAS Number:
Molecular Weight:
506.20 (free acid basis)
Beilstein:
6047109
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

natural (organic)

Assay

≥93% (HPLC)

form

powder

color

white

solubility

H2O: 50 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].[H]O[H].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)NP([O-])([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H17N6O12P3/c11-8-5-9(13-2-12-8)16(3-14-5)10-7(18)6(17)4(27-10)1-26-31(24,25)28-30(22,23)15-29(19,20)21/h2-4,6-7,10,17-18H,1H2,(H,24,25)(H2,11,12,13)(H4,15,19,20,21,22,23)/t4-,6-,7-,10-/m1/s1

InChI key

PVKSNHVPLWYQGJ-KQYNXXCUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate has been used in a study to research the effects of monovalent cations Li+, Na+, K+, NH4+, Rb+ and Cs+ on the solid and solution structures of the nucleic acid components. Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate has also been used in a study to determine that antidepressants inhibit interferon-γ-induced microglial production of IL-6 and nitric oxide.

Biochem/physiol Actions

A non-hydrolyzable ATP analog. AMP-PNP competitively inhibits ATP-dependent enzyme systems, such as glutamine synthetase. It inhibits a number of enzymes which require the hydrolysis of ATP such as DNA topoisomerase II, SV40 large T-antigen helicase and a number of kinases. Blocks ATP-sensitive calcium-dependent potassium channels.

Caution

AMP-PNP is very unstable in acidic conditions; rapidly hydrolyzes to the phosphoramidate and inorganic phosphate.

Reconstitution

A 10 mg/mL stock solution may be made, aliquoted and stored at -70 °C for up to 3 months.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Gayathri et al.
Science (New York, N.Y.), 338(6112), 1334-1337 (2012-11-01)
To ensure their stable inheritance by daughter cells during cell division, bacterial low-copy-number plasmids make simple DNA segregating machines that use an elongating protein filament between sister plasmids. In the ParMRC system of the Escherichia coli R1 plasmid, ParM, an
Vladimir M Korkhov et al.
Nature, 490(7420), 367-372 (2012-09-25)
The ATP-binding cassette (ABC) transporter BtuCD mediates the uptake of vitamin B(12) across the inner membrane of Escherichia coli. Previous structures have shown the conformations of apo states, but the transport mechanism has remained unclear. Here we report the 3.5 Å
Timothy O Street et al.
Molecular cell, 42(1), 96-105 (2011-04-09)
Hsp90 is a ubiquitous molecular chaperone. Previous structural analysis demonstrated that Hsp90 can adopt a large number of structurally distinct conformations; however, the functional role of this flexibility is not understood. Here we investigate the structural consequences of substrate binding
Maria Chiara Monti et al.
Nucleic acids research, 39(18), 8052-8064 (2011-07-09)
The DNA mismatch repair protein MutS recognizes mispaired bases in DNA and initiates repair in an ATP-dependent manner. Understanding of the allosteric coupling between DNA mismatch recognition and two asymmetric nucleotide binding sites at opposing sides of the MutS dimer
Ken-ichi Miyazono et al.
Journal of bacteriology, 194(3), 607-616 (2011-11-22)
Carbon catabolite repression (CCR) is a widespread phenomenon in many bacteria that is defined as the repression of catabolic enzyme activities for an unfavorable carbon source by the presence of a preferable carbon source. In Streptomyces, secondary metabolite production often

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service