Skip to Content
Merck
All Photos(2)

Documents

C8031

Sigma-Aldrich

N6-Cyclopentyladenosine

solid

Synonym(s):

CPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O4
CAS Number:
Molecular Weight:
335.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

white to off-white

solubility

H2O: 1.7 mg/mL
0.1 M HCl: 12 mg/mL
DMSO: 23 mg/mL

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NC4CCCC4)ncnc23

InChI

1S/C15H21N5O4/c21-5-9-11(22)12(23)15(24-9)20-7-18-10-13(16-6-17-14(10)20)19-8-3-1-2-4-8/h6-9,11-12,15,21-23H,1-5H2,(H,16,17,19)/t9-,11-,12-,15-/m1/s1

InChI key

SQMWSBKSHWARHU-SDBHATRESA-N

Looking for similar products? Visit Product Comparison Guide

Application

N6-Cyclopentyladenosine has also been used to study its effect on p53 and caspase 3, 8, and 9 expression and apoptosis rate in MCF-7 breast cancer cell line.
N6-Cyclopentyladenosine has been used to study its effect on the cavernosal smooth muscle cells from lean and db/db (obesity and type II diabetes caused by a leptin receptor mutation) mice.

Biochem/physiol Actions

N6-Cyclopentyladenosine (CPA) is an adenosine derivative and a potent A1 adenosine receptor agonist. It acts as an anticonvulsant and might exhibit protective actions against aminophylline (AMPH)-induced seizures.
N6-Cyclopentyladenosine also exhibits analgesic effect against acute, arthritis-induced and neuropathic pain.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Physical form

hygroscopic solid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ainhoa Bilbao et al.
Addiction biology, 24(5), 1008-1018 (2019-06-27)
The co-occurrence of chronic pain and alcohol use disorders (AUDs) involves complex interactions between genetic and neurophysiological aspects, and the research has reported mixed findings when they both co-occur. There is also an indication of a gender-dependent effect; males are
Yahaira Naaldijk et al.
BMC biotechnology, 16(1), 85-85 (2016-12-03)
Preservation of human skin fibroblasts and keratinocytes is essential for the creation of skin tissue banks. For successful cryopreservation of cells, selection of an appropriate cryoprotectant agent (CPA) is imperative. The aim of this study was to identify CPAs that
Cheng-Hui Hsiao et al.
Biomedical chromatography : BMC, 33(9), e4518-e4518 (2019-02-26)
The prominent stromal compartment surrounds pancreatic ductal adenocarcinoma and protects the tumor cells from chemo- or radiotherapy. We hypothesized that our nano formulation carrying cyclopamine (CPA, stroma modulator) and paclitaxel (PTX, antitumor agent) could increase the permeation of PTX through
Claudia Iasillo et al.
Nucleic acids research, 45(17), 10229-10241 (2017-10-04)
Termination of transcription is important for establishing gene punctuation marks. It is also critical for suppressing many of the pervasive transcription events occurring throughout eukaryotic genomes and coupling their RNA products to efficient decay. In human cells, the ARS2 protein
Individual and combined effects of N6-cyclopentyladenosine, flunarizine and diazepam on aminophylline-induced recurrent generalized seizures in mice
Jaishree J, et al.
Polish Journal of Pharmacology, 55(4), 559-564 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service