Skip to Content
Merck

E2387

NECA

≥98% (TLC), powder, adenosine receptor agonist

Synonym(s):

NECA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H16N6O4
CAS Number:
Molecular Weight:
308.29
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5′-(N-Ethylcarboxamido)adenosine, powder

form

powder

Quality Segment

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.2 mg/mL (Solutions may be stored for several days at 4 °C.), DMSO: 14 mg/mL (Solutions may be stored for several days at 4 °C.), 0.1 M HCl: 3.4 mg/mL (Solutions may be stored for several days at 4 °C.), H2O: insoluble, ethanol: soluble (Solutions may be stored for several days at 4 °C.)

storage temp.

2-8°C

SMILES string

CCNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23

InChI

1S/C12H16N6O4/c1-2-14-11(21)8-6(19)7(20)12(22-8)18-4-17-5-9(13)15-3-16-10(5)18/h3-4,6-8,12,19-20H,2H2,1H3,(H,14,21)(H2,13,15,16)/t6-,7+,8-,12+/m0/s1

InChI key

JADDQZYHOWSFJD-FLNNQWSLSA-N

General description

5′-(N-Ethylcarboxamido)adenosine stimulates vasodilation in the pulmonary circulation through A2-adenosine-receptor activation.

Application

5′-(N-Ethylcarboxamido)adenosine has been used:
  • for the purification of human adenosine A2Areceptor (A2AR) in decylmaltoside (DM)
  • to determine nonspecific binding of adenosine receptors
  • to investigate adenosine 2B receptor (RA2B) mechanism of action by competition assay

Biochem/physiol Actions

Potent adenosine receptor agonist with nearly equal affinity at A1 and A2 receptors.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Still not finding the right product?

Explore all of our products under 5′-(N-Ethylcarboxamido)adenosine


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
E2387-50MG04061833602607