Skip to Content
Merck
All Photos(2)

Key Documents

A0542

Sigma-Aldrich

Amprolium hydrochloride

Synonym(s):

1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H19N4Cl · HCl
CAS Number:
Molecular Weight:
315.24
Beilstein:
4118699
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

cationic

Quality Level

Assay

≥98.0% (TLC)

form

powder

mol wt

315.24 g/mol

technique(s)

cell culture | mammalian: suitable
microbiological culture: suitable

SMILES string

[Cl-].Cl[H].CCCc1ncc(C[n+]2ccccc2C)c(N)n1

InChI

1S/C14H19N4.2ClH/c1-3-6-13-16-9-12(14(15)17-13)10-18-8-5-4-7-11(18)2;;/h4-5,7-9H,3,6,10H2,1-2H3,(H2,15,16,17);2*1H/q+1;;/p-1

InChI key

PJBQYZZKGNOKNJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Bizon-Zygmańska et al.
Neurochemistry international, 59(2), 208-216 (2011-06-16)
Inhibition of pyruvate (PDHC) and ketoglutarate (KDHC) dehydrogenase complexes induced by thiamine pyrophosphate deficits is known cause of disturbances of cholinergic transmission in the brain, yielding clinical symptoms of cognitive, vegetative and motor deficits. However, particular alterations in distribution of
Gabrielle Young et al.
Veterinary parasitology, 178(3-4), 346-349 (2011-02-22)
This study evaluated the efficacy of two different doses of amprolium in goats heavily infected with pathogenic Eimeria species. Forty Boer goat kids ranging from 3 to 5 months of age with naturally occurring coccidiosis were randomly divided into 2
N E Nweze et al.
Journal of ethnopharmacology, 122(1), 6-9 (2008-12-17)
Ageratum conyzoides (Asteraceae) is folklorically used to treat diseases associated with bleeding. A study was conducted to determine the efficacy of Ageratum conyzoides in treating caecal coccidiosis of broilers. Acute toxicity test was done using thirty 28 days old broiler
Clara Lemos et al.
European journal of pharmacology, 682(1-3), 37-42 (2012-03-06)
The aim of this study was to characterize the intestinal absorption of thiamine, by investigating the hypothesis of an involvement of Organic Cation Transporter (OCT) family members in this process. [(3)H]-T(+) uptake was found to be: 1) time-dependent, 2) Na(+)-
Studies on the Eimeria of goats at Magadu Dairy Farm SUA, Morogoro, Tanzania.
E N Kimbita et al.
Tropical animal health and production, 41(7), 1263-1265 (2009-02-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service