Skip to Content
Merck
All Photos(1)

Key Documents

88571

Sigma-Aldrich

1,6-Hexanediol solution

BioUltra, ~6 M in H2O

Synonym(s):

α,ω-Hexanediol, Hexamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

form

liquid

concentration

~6 M in H2O

impurities

insoluble matter, passes filter test

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.1
λ: 280 nm Amax: 0.1

SMILES string

OCCCCCCO

InChI

1S/C6H14O2/c7-5-3-1-2-4-6-8/h7-8H,1-6H2

InChI key

XXMIOPMDWAUFGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nataliya Shulga et al.
Molecular and cellular biology, 23(2), 534-542 (2003-01-02)
The nuclear pore complex (NPC) is a permeable sieve that can dilate to facilitate the bidirectional translocation of a wide size range of receptor-cargo complexes. The binding of receptors to FG nucleoporin docking sites triggers channel gating by an unknown
D Pandolfi et al.
Nucleosides & nucleotides, 18(9), 2051-2069 (1999-11-05)
Synthetic oligonucleotides are increasingly used because of their potential activity as regulators of gene expression. One of their major drawbacks is instability toward nucleases, in particular exonucleases. In this article, we studied some terminal modifications that can enhance exonuclease resistance
M A Rahman et al.
Dental materials journal, 14(2), 230-235 (1995-12-01)
The optimum concentration and application time for an experimental dentin primer composed of an aqueous solution of 1,6-hexanediol were determined by measuring the wall-to-wall polymerization contraction gap width of a commercial light-activated resin composite in a cylindrical dentin cavity prepared
M J Politis et al.
Journal of neurocytology, 9(4), 505-516 (1980-08-01)
This study examined the thesis that 2,5-dexanedione (2,5-HD) produces distal (dying-back) axonopathy by direct toxic action on nerve fibers. Single or repeated application of undiluted or 10% 2,5-HD to exposed rat sciatic nerves caused some fibres to develop focal axonal
Jennifer Tuma et al.
The journal of physical chemistry. B, 111(45), 13101-13106 (2007-10-26)
The solution structure of a synthetic DNA mini-hairpin possessing a stilbenediether linker and three G:C base pairs has been obtained using (1)H NMR spectral data and constrained torsion angle molecular dynamics. Notable features of this structure include a compact hairpin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service