Skip to Content
Merck
All Photos(5)

Key Documents

A4382

Sigma-Aldrich

4-Aminoantipyrine

reagent grade

Synonym(s):

4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Beilstein:
181635
EC Number:
MDL number:
UNSPSC Code:
51384603
PubChem Substance ID:
NACRES:
NA.31
Pricing and availability is not currently available.

grade

reagent grade

Assay

≥97% (HPLC)

form

powder

mp

105-110 °C (lit.)

solubility

water: 50 mg/mL

SMILES string

CN1N(c2ccccc2)C(=O)C(N)=C1C

InChI

1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3

InChI key

RLFWWDJHLFCNIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reagent for glucose determination in the presence of phenol and peroxidase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Z El-Sonbati et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 213-221 (2012-12-27)
A novel series of copper(II) and palladium(II) with 4-derivatives benzaldehyde pyrazolone (L(n)) were synthesized. The mixed ligand complexes were prepared by using 1,10-phenanthroline (Phen) as second ligand. The structure of these complexes was identified and confirm by elemental analysis, molar
Yue Teng et al.
Journal of hazardous materials, 190(1-3), 574-581 (2011-04-19)
4-Aminoantipyrine (AAP) is widely used in the pharmaceutical industry, in biochemical experiments and in environmental monitoring. AAP as an aromatic pollutant in the environment poses a great threat to human health. To evaluate the toxicity of AAP at the protein
Isela Lavilla et al.
Talanta, 98, 197-202 (2012-09-04)
In this work, a new method based on the combination of dispersive liquid-liquid microextraction (DLLME) with microvolume spectrophotometry has been developed as a greener and miniaturized alternative to the 5530 APHA standard method for determining phenols in water and wastewater.
N Raman et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 85(1), 223-234 (2011-10-25)
Few novel 4-aminoantipyrine derived Schiff bases and their metal complexes were synthesized and characterized. Their structural features and other properties were deduced from the elemental analysis, magnetic susceptibility and molar conductivity as well as from mass, IR, UV-vis, (1)H NMR
Qihui Wang et al.
Journal of hazardous materials, 186(2-3), 1076-1081 (2010-12-21)
In this work, the immobilization of 4-aminoantipyrine onto bentonite was carried out and it was then used to investigate the adsorption behavior of Cr(III), Hg(II) and Pb(II) ions from aqueous solutions. The separation and preconcentration conditions of analytes were investigated

Protocols

Enzymatic assay of lipase type XIII from Pseudomonas sp. using a coupled enzyme system of glycerol kinase and glycerophosphate oxidase (EC 3.1.1.3)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the recommended storage method for stock solutions of item A4382, 4-Aminoantipyrine?

    1 answer
    1. To enhance solubility, it is advised to warm the tube at 37 ℃ and subsequently shake it in an ultrasonic bath for a brief period. Stock solutions can be stored at temperatures below -20℃ for several months. Ideally, it is recommended to prepare and utilize the solution on the same day. However, if advance preparation is necessary, stock solutions must be sealed and stored below -20℃. Prior to use, allowing the vial to reach room temperature for at least an hour is recommended.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service