Skip to Content
Merck
All Photos(1)

Key Documents

T7883

Sigma-Aldrich

Trimethoprim

≥98.5%

Synonym(s):

2,4-Diamino-5-(3,4,5-trimethoxybenzyl)pyrimidine, NSC 106568

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N4O3
CAS Number:
Molecular Weight:
290.32
Beilstein:
625127
EC Number:
MDL number:
UNSPSC Code:
51285203
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥98.5%

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.)

color

white to light yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

COc1cc(Cc2cnc(N)nc2N)cc(OC)c1OC

InChI

1S/C14H18N4O3/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-17-14(16)18-13(9)15/h5-7H,4H2,1-3H3,(H4,15,16,17,18)

InChI key

IEDVJHCEMCRBQM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: pyrimidine

Application

Primarily used as an antibacterial agent. Dihydrofolate reductase inhibitor with selectivity for the prokaryote enzyme.
Trimethoprim is primarily used as an antibacterial agent. It has dihydrofolate reductase inhibitor activity with selectivity for the prokaryote enzyme. It is used to treat urinary tract infections, mild acute prostatitis, recurrent cystitis, asymptomatic bacteriuria during pregnancy and respiratory tract infections.

Biochem/physiol Actions

The combination of trimethoprim and sulfamethoxazole interferes with the cellular metabolism of folic acid in the bacterial cell by blocking the biosynthesis of nucleotides. Trimethoprim binds to dihydrofolate reductase and inhibits the reduction of dihydrofolic acid (DHF) to tetrahydrofolic acid (THF). THF is an essential precursor in the thymidine synthesis pathway and interference with this pathway inhibits bacterial DNA synthesis.

Packaging

5G,25G,100G

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Gilad et al.
Nature communications, 11(1), 1744-1744 (2020-04-10)
Association areas in neocortex encode novel stimulus-outcome relationships, but the principles of their engagement during task learning remain elusive. Using chronic wide-field calcium imaging, we reveal two phases of spatiotemporal refinement of layer 2/3 cortical activity in mice learning whisker-based
Mira Rakic-Martinez et al.
Applied and environmental microbiology, 77(24), 8714-8721 (2011-10-18)
Listeria monocytogenes is a leading agent for severe food-borne illness and death in the United States and other nations. Even though drug resistance has not yet threatened therapeutic interventions for listeriosis, selective pressure associated with exposure to antibiotics and disinfectants
Kate S Baker et al.
The Lancet. Infectious diseases, 15(8), 913-921 (2015-05-06)
Shigellosis is an acute, severe bacterial colitis that, in high-income countries, is typically associated with travel to high-risk regions (Africa, Asia, and Latin America). Since the 1970s, shigellosis has also been reported as a sexually transmitted infection in men who
Crystal Y Chen et al.
PLoS pathogens, 5(4), e1000392-e1000392 (2009-04-22)
The role of CD8 T cells in anti-tuberculosis immunity in humans remains unknown, and studies of CD8 T cell-mediated protection against tuberculosis in mice have yielded controversial results. Unlike mice, humans and nonhuman primates share a number of important features
Kyuho Han et al.
Nature methods, 11(1), 86-93 (2013-11-12)
Protein concentrations are often regulated by dynamic changes in translation rates. Nevertheless, it has been challenging to directly monitor changes in translation in living cells. We have developed a reporter system to measure real-time changes of translation rates in human

Articles

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

Biofiles reviews innovative technologies for cancer research, reflecting the complexity of the disease.

Cancer research innovations address the complexity of the disease, providing advanced technologies for varied applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service