Skip to Content
Merck
All Photos(1)

Key Documents

51411C

SAFC

Minimum Essential Medium

with Earle′s Balanced Salts, with 2.0 mM L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Minimum Essential Medium Eagle, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

Earle’s salts (5% CO2): yes
phenol red: 10.62 mg/L
NaHCO3: 2200 mg/L
L-glutamine: 290 mg/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium (MEM) with Earle′s Balanced Salts was developed by Harry Eagle as a result of his studies to determine the essential nutritional requirements of mouse L cells and HeLa cells in culture. He demonstrated that these cells require 13 amino acids and seven vitamins to grow and reproduce in vitro . MEM is a modification of Eagle′s earlier medium Basal Medium Eagle (BME), containing higher concentrations of the essential nutrients. These media have demonstrated the ability to support a variety of normal and transformed cells in culture and contain Earle′s Balanced Salts, which make them suitable for use in atmospheres charged with CO2.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dariusz Dąbruś et al.
International journal of molecular sciences, 21(11) (2020-06-14)
This research aimed to assess the impact of cisplatin, depending on the concentration and exposure time, on the expression pattern of leptin in an endometrial cancer cell line. Ishikawa endometrial cancer cell cultures were incubated with cisplatin, at concentrations of
Robert Nason et al.
Journal of the Association for Research in Otolaryngology : JARO, 10(2), 151-160 (2009-01-16)
Osteoclasts are the only cells capable of carrying out bone resorption and therefore are responsible for the osteolysis seen in infectious diseases such as chronic otitis media and infected cholesteatoma. Pseudomonas aeruginosa is the most common organism isolated from these
Wenxin Wu et al.
Virology, 396(2), 178-188 (2009-11-17)
We studied cytokine responses to influenza virus PR8 (H1N1) and Oklahoma/309/06 (OK/06, H3N2) in a novel human lung tissue model. Exposure of the model to influenza virus rapidly activated the mitogen-activated protein kinase signaling (MAPK) pathways ERK, p38 and JNK.
Chia-Chyi Liu et al.
PloS one, 3(3), e1810-e1810 (2008-03-20)
This work investigated the replication kinetics of the four dengue virus serotypes (DEN-1 to DEN-4), including dengue virus type 4 (DEN-4) recovered from an infectious cDNA clone, in Vero cells and in MRC-5 cells grown on Cytodex 1 microcarriers. DEN-1
Petra G Hirrlinger et al.
PloS one, 6(1), e16106-e16106 (2011-02-02)
Glial cells such as retinal Müller glial cells are involved in potassium ion and water homeostasis of the neural tissue. In these cells, inwardly rectifying potassium (Kir) channels and aquaporin-4 water channels play an important role in the process of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service