Skip to Content
Merck
All Photos(1)

Key Documents

M7145

Sigma-Aldrich

MEM Non-essential Amino Acid Solution (100×)

Without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

MEM NEAA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160406
NACRES:
NA.75

Product Name

MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

General description

Eagle′s minimum essential medium (MEM) is a synthetic culture medium, which is composed of balanced salt solutions, and amino acids and vitamins. These components are essential for the growth of cultured cells. MEM might be supplemented with non-essential amino acids, to make MEM non-essential amino acid solution. The non-essential amino acids include L-alanine, L-asparagine, L-aspartic acid, L-glycine, L-serine, L-proline and L-glutamic acid.

Application

Adding supplements of amino acids to media both stimulates growth and prolongs the viability of cells in culture. MEM Non-essential Amino Acid Solution supplement adds nutrients and reduces the biosynthetic burden on cell in vitro.
MEM Non-essential Amino Acid Solution has been used in Dulbecco′s minimum essential media for the maintenance of mammalian cells. It has been supplemented in medium prepared for oocyte maturation.

Biochem/physiol Actions

Studies on hybridoma culture shows that addition of Eagle′s minimum essential medium (MEM) non-essential amino acid leads to enhanced membrane stability, during death phase of the culture. This also results in increased antibody secretion during death phase, and decreased lactate concentration.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell Culture
Jakoby WB
Methods in Enzymology, 58 (1979)
Influence of non-essential amino acids on apoptotic and necrotic death of mouse hybridoma cells in batch cultures
Fassnacht D
Biotechnology Letters, 19(1), 35-38 (1997)
Ayako Suzuki et al.
Genome biology, 16, 66-66 (2015-04-19)
To understand the heterogeneous behaviors of individual cancer cells, it is essential to investigate gene expression levels as well as their divergence between different individual cells. Recent advances in next-generation sequencing-related technologies have enabled us to conduct a single-cell RNA-Seq
Excessive concentration of glucose during in vitro maturation impairs the developmental competence of bovine oocytes after in vitro fertilization: relevance to intracellular reactive oxygen species and glutathione contents.
Hashimoto S et al.
Molecular Reproduction and Development, 56, 520-520 (2000)
Janis A Müller et al.
Medical microbiology and immunology, 206(2), 175-185 (2017-02-09)
Zika virus (ZIKV) is an emerging pathogen that causes congenital infections which may result in birth defects, such as microcephaly. Currently, no approved treatment or vaccination is available. ZIKV can be readily detected in cell culture where virally infected cells

Articles

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Protocols

This detailed procedure allows you to transduce Mouse Embryonic Fibroblasts (MEF) using MISSION ExpressMag Super Magnetic Kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service