Skip to Content
Merck
All Photos(4)

Key Documents

223387

Sigma-Aldrich

Nickel(II) chloride hexahydrate

ReagentPlus®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2 · 6H2O
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
powder or crystals
Pricing and availability is not currently available.

grade

reagent

Quality Level

product line

ReagentPlus®

form

powder or crystals

concentration

23.0-26.0% Ni (EDTA titration)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ni]Cl

InChI

1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LAIZPRYFQUWUBN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nickel(II) chloride hexahydrate is a hydrated nickel salt. Its crystals were monoclinc and crystal structure was investigated by X-ray single crystal method.[1]

Application

Nickel(II) chloride hexahydrate may be employed as a catalyst in the high yield synthesis of tetra-substituted pyrrole derivatives.[2] It may be used for the conversion of tetrahydropyranyl (THP) and tert-butyldimethylsilyl (TBS) ethers to the corresponding hydroxyl compounds.[3]
Nickel(II) chloride hexahydrate may be used as a catalyst in the following processes:
  • Synthesis of tetra-substituted pyrrole derivatives via four-component condensation reaction of aromatic aldehydes, benzylamines, β-ketoesters and nitroalkanes.[2]
  • In combination with 1,2-ethanedithiol for the deprotection of tetrahydropyranyl (THP) and tert-butyldimethylsilyl (TBS) ethers to their corresponding hydroxyl compounds.[3]
  • Synthesis of 3,4-dihydropyrimidinones from aldehydes and β-keto esters via Biginelli condensation.[4]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysis of the Biginelli reaction by ferric and nickel chloride hexahydrates. One-pot synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones.
Lu J and Bai Y.
Synthesis, 2002(04), 0466-0470 (2002)
The Crystal Structure of Nickel Chloride Hexahydrate, NiCl2.6H2O.
Mizuno J.
Journal of the Physical Society of Japan, 16(8), 1571-1580 (1961)
Synthesis of tetra-substituted pyrroles, a potential phosphodiesterase 4B inhibitor, through nickel (II) chloride hexahydrate catalyzed one-pot four-component reaction.
Khan AT, et al.
Tetrahedron Letters, 53(32), 4145-4150 (2012)
A catalytic amount of nickel (II) chloride hexahydrate and 1, 2-ethanedithiol is a good combination for the cleavage of tetrahydropyranyl (THP) and tert-butyldimethylsilyl (TBS) ethers.
Khan AT, et al.
Tetrahedron Letters, 45(52), 9617-9621 (2004)
Jenilee Way et al.
Nuclear medicine and biology, 40(3), 430-436 (2013-01-12)
4-[(18)F]Fluorobenzylamine ([(18)F]FBA) is an important building block for the synthesis of (18)F-labeled compounds. Synthesis of [(18)F]FBA usually involves application of strong reducing agents like LiAlH4 which is challenging to handle in automated synthesis units (ASUs). Therefore, alternative methods for the

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service