Skip to Content
Merck
All Photos(4)

Key Documents

255599

Sigma-Aldrich

Cobalt(II) chloride hexahydrate

ACS reagent, 98%

Synonym(s):

Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

vapor pressure

40 mmHg ( 0 °C)

Assay

98%
98.0-102.0% (ACS specification)

form

powder, crystals or chunks

reaction suitability

reagent type: catalyst
core: cobalt

impurities

≤0.01% insolubles

anion traces

nitrate (NO3-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.005%
K: ≤0.01%
Mg: ≤0.005%
Na: ≤0.05%
Ni: ≤0.1%
Zn: ≤0.03%

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II) chloride hexahydrate, also known as cobaltous chloride hexahydrate, is a hydrated form of cobalt and chloride. It is a crystalline solid consisting of octahedral complexes, with the coordination polyhedron of the Co2+ ion including four H2O molecules and two Cl ions. It is highly soluble in water and is commonly used as a catalyst in various organic synthesis, such as during the synthesis of benzimidazole and biscoumarin derivatives.

Application

Cobalt(II) chloride hexahydrate can be used as a catalyst for the:
  • Dioxygenation of styrenes with N-hydroxyphthalimide to synthesize corresponding alcohols and ketones.
  • Oxidative coupling of primary amines to imines.
  • Friedel-Crafts reaction between indoles and activated olefins to synthesize β-indolylketones.
  • Synthesis of β-enaminones and β-enamino esters.
  • Reaction of acyl chlorides with diselenides or disulfides to synthesize selenol and thiol esters.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CoCl2. 6H2O crystal growth and its spectral characteristics
Diana Sergeevna M, et al.
Optical Materials, 144, 114339-114339 (2023)
Cobalt (II) Chloride Hexahydrate as an Efficient and Inexpensive Catalyst for the Preparation of Biscoumarin Derivatives.
Mohammad Reza N, et al.
Advances in Chemistry, 1-6 null
A simple and convenient one-pot synthesis of benzimidazole derivatives using cobalt (II) chloride hexahydrate as catalyst
Abu K T, et al.
Synthetic Communications, 39, 2339-2346 (2009)
Preparation of PVDF nanofiber composites for hydrogen generation from sodium borohydride.
Chinnappan A, et al.
Energy, 36(2), 755-759 (2011)
A simple and convenient one-pot synthesis of benzimidazole derivatives using cobalt (II) chloride hexahydrate as catalyst.
Khan AT, et al.
Synthetic Communications, 39(13), 2339-2346 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service