Skip to Content
Merck
All Photos(4)

Documents

203874

Sigma-Aldrich

Nickel(II) nitrate hexahydrate

99.999% trace metals basis

Synonym(s):

Nickel dinitrate hexahydrate, Nickel nitrate hexahydrate, Nickel(2+) dinitrate hexahydrate, Nickelous nitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
290.79
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

solid

impurities

≤15.0 ppm Trace Metal Analysis

mp

56 °C (lit.)

density

2.05 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Ni.6H2O/c2*2-1(3)4;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;

InChI key

AOPCKOPZYFFEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nickel(II) nitrate hexahydrate can be used:

  • As an additive in the fabrication of cellulose acetate polymers(CA) to control the porosity of the polymer. The formation of Ni(NO3)2.6H2O aggregates in polymer matrix during solidification and the strong interaction between Ni, nitrate, and water molecules result in the formation of well-defined pores on the surface of the CA matrix.
  • As a starting material to prepare nickel(ii) Schiff base, which is used as a precursor to synthesize NiO nanoparticles by solid-state thermal decomposition method.
  • As a dopant to prepare Ni-Ceria catalyst for selective hydrogenation of acetylene.
  • As a catalyst to prepare 3,4-dihydropyrimidinone derivatives via Biginelli cyclocondensation.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doron Levin et al.
Inorganic chemistry, 35(14), 4191-4197 (1996-07-03)
A layered ammonium nickel molybdate was prepared by precipitation from a solution of nickel nitrate and ammonium heptamolybdate. The compound obtained, (NH(4))HNi(2)(OH)(2)(MoO(4))(2), is trigonal with hexagonal unit cell parameters a = 6.0147(4) Å, c = 21.8812(13) Å, and Z =
H Tanojo et al.
Acta dermato-venereologica. Supplementum, (212)(212), 19-23 (2002-01-23)
Allergic contact dermatitis due to nickel salts is common. It is therefore important to measure the permeation of these salts through the stratum corneum (SC), the primary rate-limiting domain in skin. An advanced diffusion system and analytical techniques now enable
Jun Li et al.
Journal of colloid and interface science, 332(1), 231-236 (2009-01-16)
In this paper, we report the successful synthesis of Ni(12)P(5) hollow spheres via a facile hydrothermal route, employing white phosphorus (WP) and nickel nitrate as the reactants in the presence of hexamethylenetetramine (HMT) and polyethylene glycol 10000 (PEG-10000). The phase
Marisa Freitas et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 26(1), 13-21 (2012-10-26)
Nickel is an ubiquitous transition metal that is industrially applied in many forms, which inevitably leads to a high degree of occupational and environmental exposure. Over-exposure to nickel can produce a variety of adverse effects on human health, including allergy
B Vikhoff et al.
Acta radiologica (Stockholm, Sweden : 1987), 36(3), 323-325 (1995-05-01)
An investigation regarding possible artefacts from dental filling materials in MR imaging is presented including 9 types of such materials from various manufacturers. Freshly extracted teeth were prepared and the filling materials were handled according to manufacturers' instructions. The teeth

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service