Skip to Content
Merck
All Photos(1)

Key Documents

451193

Sigma-Aldrich

Nickel(II) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Nickel chloride, Nickel dichloride, Nickel dichloride (NiCl2 ), Nickelous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2
CAS Number:
Molecular Weight:
129.60
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

density

3.55 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Ni]Cl

InChI

1S/2ClH.Ni/h2*1H;/q;;+2/p-2

InChI key

QMMRZOWCJAIUJA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) chloride anhydrous, powder, 99.99% trace metals basis comes in a powder form or in chunks with trace metal impurities ≤ 150.0 ppm. It is a highly useful inorganic compound with a broad range of applications across various industries. Its relatively high purity makes it suitable for both industrial and laboratory settings. It is widely used in catalysis, electroplating, battery manufacturing, chemical synthesis, material science, dye and pigment production, and laboratory research.

Application

Nickel(II) chloride anhydrous can be used in advanced studies in battery technologies, such as nickel-based cathode materials for lithium-ion batteries and alternative battery chemistries. Used to investigate and improve the properties of electrode materials in nickel-cadmium (NiCd) and nickel-metal hydride (NiMH) batteries. Research into electrocatalysts for energy applications, including water splitting and fuel cells, frequently utilizes high-purity Nickel(II) chloride to ensure optimal performance and reproducibility. It can be used as a precursor in the synthesis of nickel nanoparticles, which have applications in catalysis, magnetic materials, and electronic devices. The high purity Nickel(II) chloride serves as a precursor for various nickel compounds, such as nickel oxides and hydroxides, which are studied for their electrical, magnetic, and catalytic properties.[1][2][3]

Features and Benefits

Its high purity guarantees accurate and reproducible experimental results, making it essential for applications in catalysis, nanomaterial synthesis, battery research, material science, coordination chemistry, environmental chemistry, and analytical chemistry. The vapor-phase co-reductions with other metal halides such as aluminum chloride (cat. no. 449598) by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
The vapor-phase co-reductions with other metal halides such as aluminum chloride (cat. no. 449598) by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Hasnain Jalal et al.
Biosensors & bioelectronics, 87, 522-530 (2016-09-07)
Multimodal electrochemical technique incorporating both open circuit potential (OCP) and amperometric techniques have been conceptualized and implemented to improve the detection of specific analyte in systems where more than one analyte is present. This approach has been demonstrated through the
Samuel Dessources et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(23), 3964-3973 (2016-09-23)
Hydrogen oxidation and evolution reactions (HOR and HER) are studied on Ptx Ni1-x /C materials synthesized by the bromide anion exchange method. Physicochemical characterization shows that this surfactant-free method enables the preparation of well-dispersed and effective catalysts for the processes
Yuhang Li et al.
Nature communications, 11(1), 6190-6190 (2020-12-05)
Electroreduction uses renewable energy to upgrade carbon dioxide to value-added chemicals and fuels. Renewable methane synthesized using such a route stands to be readily deployed using existing infrastructure for the distribution and utilization of natural gas. Here we design a
Tamzin A Blewett et al.
Archives of environmental contamination and toxicology, 68(2), 382-394 (2014-12-30)
The mechanisms of nickel (Ni) toxicity in marine fish remain unclear, although evidence from freshwater (FW) fish suggests that Ni can act as a pro-oxidant. This study investigated the oxidative stress effects of Ni on the euryhaline killifish (Fundulus heteroclitus)
Katherine R Brimblecombe et al.
ACS chemical neuroscience, 6(1), 124-129 (2014-12-02)
Neuronal T-type voltage-gated Ca(2+) channels are reported to have physiological roles that include regulation of burst firing, Ca(2+) oscillations, and neurotransmitter release. These roles are often exposed experimentally by blocking T-type channels with micromolar Ni(2+). We used Ni(2+) to explore

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service