Skip to Content
Merck
All Photos(3)

Key Documents

N6136

Sigma-Aldrich

Nickel(II) chloride hexahydrate

BioReagent, suitable for cell culture

Synonym(s):

NiCl2.6H2O, Nickel chloride hexahydrate, Nickel dichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiCl2 · 6H2O
CAS Number:
Molecular Weight:
237.69
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
grade:
reagent
form:
powder

grade

reagent

product line

BioReagent

form

powder

technique(s)

cell culture | mammalian: suitable

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Ni]Cl

InChI

1S/2ClH.Ni.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

LAIZPRYFQUWUBN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Nickel(II) chloride hexahydrate has been used:
  • in the preparation of nickel platting solution for galvanostatically coating nickel inside the reduced carbon layer during the synthesis of multiwall carbon nanotube micromotors
  • in screening of metals for inflammatory effect on vascular endothelial cells
  • as an additive in artificial cerebrospinal fluid (ACSF) to analyse excitatory post synaptic currents (EPSPs) in the hippocampus slices

Biochem/physiol Actions

Nickel (Ni) is an essential trace element that plays an important role in the structure and function of DNA, RNA and protein. It has toxic effects on renal, pulmonary and cardiovascular system at higher concentrations. Nickel is implicated in carcinogenesis. Ni2+ ions is a clastogenic agent. Nickel ions (Ni2+) is a non-specific voltage-gated calcium channel blocker.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional coupling of Cav2.3 and BK potassium channels regulates action potential repolarization and short-term plasticity in the mouse hippocampus
Gutzmann JJ, et al.
Frontiers in Cellular Neuroscience, 13(3), 492-499 (2019)
Surfactant-Free beta-Galactosidase Micromotors for ?On-The-Move? Lactose Hydrolysis
Maria-Hormigos R, et al.
Advances in Functional Materials, 28(25), 1704256-1704256 (2018)
Genetic and epigenetic mechanisms in metal carcinogenesis and cocarcinogenesis: nickel, arsenic, and chromium
Salnikow K and Zhitkovich A
Chemical Research in Toxicology, 21(1), 28-44 (2007)
Studies on the genotoxic effect of nickel chloride in mice and the possible protective role of soybean seeds extracts
Fahmy MA, et al.
Global Journal of Pharmacology, 8(4), 625-634 (2014)
Nonrandom chromosomal alterations in nickel-transformed Chinese hamster embryo cells
Conway K and Costa M
Cancer Research, 49(21), 6032-6038 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service