Skip to Content
Merck

Skip To

A7906

Bovine Serum Albumin

heat shock fraction, pH 7, ≥98%

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥98%
Form:
lyophilized powder
Technique(s):
blood typing: suitable, microbiological culture: suitable
Impurities:
≤5.0% Loss on drying
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Level

assay

≥98%

form

lyophilized powder

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

blood typing: suitable, microbiological culture: suitable

impurities

≤5.0% Loss on drying

loss

≤5%

pH

7

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

BT virus, none detected, VSV virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Application

Bovine serum albumin has been used-
  • as a component of PCR (polymerase chain reaction) mix
  • in the preparation of dissociated cortical neurons
  • during immunofluorescent labeling
  • as a component of Ca2+-free saline (containing collagenase, hyaluronidase and DNAse) used for bovine tissue digestion

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Prepared using heat shock fractionation.
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3912A8022A9543
origin

USA origin

origin

USA origin

origin

USA origin

origin

USA origin

technique(s)

blood typing: suitable, microbiological culture: suitable

technique(s)

blood typing: suitable, microbiological culture: suitable

technique(s)

ELISA: suitable, blocking: suitable, electrophoresis: suitable, western blot: suitable

technique(s)

ELISA: suitable, electrophoresis: suitable, western blot: suitable

biological source

bovine

biological source

bovine

biological source

bovine

biological source

bovine

assay

≥98%

assay

≥96%

assay

≥96%

assay

≥98%

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The slice overlay assay: a versatile tool to study the influence of extracellular signals on neuronal development.
Polleux F and Ghosh A
Science's STKE : Signal Transduction Knowledge Environment, 2002(136), pl9-pl9 (2002)
V Templier et al.
Scientific reports, 7(1), 9457-9457 (2017-08-27)
Bloodstream bacterial infections are life-threatening conditions necessitating prompt medical care. Rapid pathogen identification is essential for early setting of the best anti-infectious therapy. However, the bacterial load in blood samples from patients with bacteremia is too low and under the
New tetranucleotide microsatellites for fine-scale discrimination among endangered Chinook salmon (Oncorhynchus tshawytscha).
Greig C et al
Molecular Ecology Notes, 3(3), 376-379 (2003)



Global Trade Item Number

SKUGTIN
A7906-10G04061835504510
A7906-100G04061835521043
A7906-500G04061835553631
A7906-5KG04061835515295
A7906-250G04061833693377
A7906-10KG04061835511730
A7906-1KG04061835553624
A7906-5G04061833693384
A7906-50G04061835511747

Questions

1–9 of 9 Questions  
  1. How long is BSA (A7906) viable at room temperature? 200g was left on benchtop overnight (Approximately 22 hours), will it still work?

    1 answer
    1. This item is stored refrigerated at 2-8°C. However, the product is shipped at ambient temperature. Shipping routes are configured to minimize transit time.
      Refrigerated products that ship at ambient temperature are considered stable up to 2 weeks for transit time.

      Helpful?

  2. Is the product A7906, Albumin from bovine serum (BSA), fatty acid-free?

    1 answer
    1. Helpful?

  3. After reconstitution in PBS, for how long is the protein stable in solution if stored at 4°C or -20*C?

    1 answer
    1. Prepared BSA solution products also available for purchase. These products have a shelf life ranging from 18 months to 2 years. Many are assigned a restest schedule with no firm expiration date. These products are prepared in saline, DPBS, or Tris. These solutions are filtered products and offered in a range of concentrations. The storage recommendation for these products is 2 - 8 deg. C. In addition, BSA is often used as a carrier protein for less stable compounds that are stored frozen. While there is no established shelf life for a BSA solution stored frozen, it is quite stable. See the link below for a sample Certificate of Analysis for one of the BSA solutions.

      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/A9576-50MLSLCL8813.pdf

      Helpful?

  4. Does this BSA have bovine immunoglobulins?

    1 answer
    1. This product does contain bovine immunoglobulins. BSA products that do not contain immunoglobulins will state "globulin-free" or "essentially globulin-free" on the product detail page. Product A3156 is an example of an immunoglobulin free BSA product.

      Helpful?

  5. Is Product A7906, Albumin from bovine serum (BSA), suitable for use as a blocking agent in ELISA?

    1 answer
    1. We have not tested the use of A7906 in ELISA. Because the remainder of the product is mostly globulins, we would not suggest A7906 for ELISA use.

      Helpful?

  6. Can Product A7906, Albumin from bovine serum (BSA), be used in immunohistochemistry?

    1 answer
    1. We have not ourselves tested A7906 for use in immunohistochemistry. However, a literature search can turn up publications that cite this use, such as this paper: Surendran, K., et al., Wnt-Dependent Beta-Catenin Signaling Is Activated after Unilateral Ureteral Obstruction, and Recombinant Secreted Frizzled-Related Protein 4 Alters the Progression of Renal Fibrosis. J. Am. Soc. Nephrol., Vol. 16, pp. 2373-2384 (2005).

      Helpful?

  7. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  8. Is Product A7906, Albumin from bovine serum (BSA), tested for insulin levels?

    1 answer
    1. A7906 is not tested in quality control for insulin levels. One BSA that is indicated as "suitable for insulin RIA procedures" is A7888.

      Helpful?

  9. Is Product A7906, Albumin from bovine serum (BSA), protease-free?

    1 answer
    1. We have not tested A7906 in quality control to be protease-free. Other BSA products to suggest that are protease-free would be A3059 or B6917.

      Helpful?

Reviews

No rating value

Active Filters