Skip to Content
Merck

A3294

Bovine Serum Albumin

heat shock fraction, protease free, pH 7, ≥98%

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥98%
Form:
lyophilized powder
Technique(s):
ELISA: suitable, blood typing: suitable, microbiological culture: suitable
Impurities:
protease, none detected
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Level

assay

≥98%

form

lyophilized powder

purified by

heat shock fractionation

packaging

poly bottle of

origin

USA origin

technique(s)

ELISA: suitable, blood typing: suitable, microbiological culture: suitable

impurities

protease, none detected

loss

≤5%

pH

7

solubility

water: soluble (40 mg/ml)

UniProt accession no.

foreign activity

BT Virus, none detected, VSV Virus, none detected

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

Albumin is abundantly present in human plasma. Increased levels of albumin result in dehydration. Albumin turnover is usually found in children with iron deficiency anemia. It is a predictive indicator for liver disease.

Application

Bovine Serum Albumin has been used:
  • to block non-specific binding of the antibodies
  • as a component of activating medium for inducing sperm motility
  • to determine the protein concentration of the bacteriocin by the bicinchoninic acid (BCA) method

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Features and Benefits

  • Heat shock fractionated
  • Protease-free

Preparation Note

Prepared using heat shock fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Cryopreservation of Atlantic cod Gadus morhua L. spermatozoa: effects of extender composition and freezing rate on sperm motility, velocity and morphology
Butts I A E, et al.
Cryobiology, 61(2), 174-181 (2010)
Histomorphometric alteration and cell-type specific modulation of arylhydrocarbon receptor and estrogen receptor expression by 2, 3, 7, 8-tetrachlorodibenzo-p-dioxin and 17beta-estradiol in mouse experimental model of endometriosis
Kitajima M, et al.
Reproductive Toxicology, 18(6), 793-801 (2004)
Leuconostoc carnosum 4010 has the potential for use as a protective culture for vacuum-packed meats: culture isolation, bacteriocin identification, and meat application experiments
Budde B B, et al.
International Journal of Food Microbiology, 83(2), 171-184 (2003)



Global Trade Item Number

SKUGTIN
A3294-10G04061835553297
A3294-100G04061835553280
A3294-5G04061833425893
A3294-50G04061833360651
A3294-500G04061833360644
A3294-1KG04061833360637
A3294-10KG04061833484029
A3294-5KG04061833458334