Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥96%
Form:
lyophilized powder
Technique(s):
ELISA: suitable, blocking: suitable, electrophoresis: suitable, western blot: suitable
Impurities:
BT and VSV virus, tested
biological source
bovine
Quality Segment
assay
≥96%
form
lyophilized powder
purified by
heat shock fractionation
packaging
poly bottle of
origin
USA origin
technique(s)
ELISA: suitable, blocking: suitable, electrophoresis: suitable, western blot: suitable
impurities
BT and VSV virus, tested
pH
5.2
solubility
water: soluble (40 mg/ml)
UniProt accession no.
foreign activity
BT virus, none detected, VSV virus, none detected
storage temp.
2-8°C
Gene Information
bovine ... ALB(280717)
General description
Bovine serum albumin (BSA) is a serum protein with a calculated molecular weight of 66,430 Daltons BSA consists of three domains with two subdomains under each, and is a α-helical, globular and non-glycosylated protein with 17-disulfide bonds.
BSA acts as a carrier protein for biomolecules like as fatty acids, amino acids, and steroids.
BSA acts as a carrier protein for biomolecules like as fatty acids, amino acids, and steroids.
Application
Bovine Serum Albumin has been used:
Although we have not specifically tested this product in the following applications, several publications have cited use of A8022 in various applications such as:
- with primary antibodies to prevent non-specific binding in immunohistochemistry of heart sections
- as a component of trypsin inhibitor solution
- as a component of sterile sorting buffer for immunocytochemistry of common lymphoid progenitor (CLP) cells
Although we have not specifically tested this product in the following applications, several publications have cited use of A8022 in various applications such as:
- Embryo culture (Dobrinsky, J.R. et al., Biol. Reprod., 55(5), 1069-1074 (1996))
- Oocyte culture medium (Jung, Y.J., and Cheon, Y.-P., Dev. Reprod., 18(2), 117-125 (2014))
- Western blotting (Chronopoulos, A. et al., Nat. Commun., 7:7, 12630 (doi: 10.1038/ncomms12630) (2016)
Biochem/physiol Actions
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Bovine Serum Albumin is used in many applications, such as the following:
- A transporter for drugs, hormones and fatty acids.
- As a blocking agent in enzyme linked immunosorbent assay (ELISA) for preventing non-specific binding of antigens and antibodies to microtiter plates
- BSA is a crucial component of cell culture media and favors embryonic stem cells (hESC) differentiation.
- BSA shares structural features with human serum albumin. Its high solubility, low cost and purity and interaction with surfactants make it useful in the cosmetic and pharmaceutical industries.
Preparation Note
Prepared using heat shock fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.
Still not finding the right product?
Explore all of our products under Bovine Serum Albumin
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A8022-500G | 04061833391044 |
| A8022-10G | 04061833391020 |
| A8022-50G | 04061833391051 |
| A8022-1KG | 04061833391037 |
| A8022-100G | 04061833391013 |
| A8022-5G | 04061833693407 |